Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Break free from the long reaction times, high costs, and method complexity of manual proteomic sample preparation with the Thermo Scientific™ AccelerOme™ automated sample preparation platform. With factory-supplied reagents and kits, and step-by-step onscreen instructions, high reproducibility and confident results are delivered for both label-free and Tandem Mass Tag (TMT) multiplexing strategies with minimal effort. The AccelerOme platform is optimized to fit into the Thermo Scientific™ Orbitrap™ mass spectrometer ecosystem, streamlining the entire workflow from sample preparation to data processing.
Description | Replacement needle |
Unit Size | Each |
Description | With tray cooling |
Includes | Detector: In-line flow-through UV spectrophotometer |
Capacity | Label Free: 36 samples per run TMT11: 33 samples per run TMTPro: 32 samples per run |
Temperature Setting | 6-40°C |
Unit Size | Each |
Description | Without tray cooling |
Includes | Detector: In-line flow-through UV spectrophotometer |
Capacity | Label Free: 36 samples per run TMT11: 33 samples per run TMTPro: 32 samples per run |
Unit Size | Each |
Description | Tray cooling module |
Temperature Setting | 6-40°C |
Unit Size | Each |
Description | Replacement 1 mL dilutor syringe |
Unit Size | Each |
Description | Periodic Maintenance kit, end user |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | Price (USD) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
C0950-01-01219 | Each | Replacement needle | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
C0960-01-00109 | Each | With tray cooling | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
C0960-01-00110 | Each | Without tray cooling | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
C0960-01-00111 | Each | Tray cooling module | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
1R77010-1136 | Each | Replacement 1 mL dilutor syringe | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
C0950-01-0013 | Each | Periodic Maintenance kit, end user | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
The AccelerOme automated sample preparation platform features include:
Operational ease of use:
Commercially available kits: