Thermo Scientific™

AccelerOme™ Automated Sample Preparation Platform

Catalog number: C0960-01-00111
Have Questions?
Thermo Scientific™

AccelerOme™ Automated Sample Preparation Platform

Catalog number: C0960-01-00111
Have Questions?

Break free from the long reaction times, high costs, and method complexity of manual proteomic sample preparation with the Thermo Scientific™ AccelerOme™ automated sample preparation platform. With factory-supplied reagents and kits, and step-by-step onscreen instructions, high reproducibility and confident results are delivered for both label-free and Tandem Mass Tag (TMT) multiplexing strategies with minimal effort. The AccelerOme platform is optimized to fit into the Thermo Scientific™ Orbitrap™ mass spectrometer ecosystem, streamlining the entire workflow from sample preparation to data processing.

 

 
Catalog Number
C0960-01-00111
Unit Size
Each
Description
Tray cooling module
Price (USD)
Full specifications
DescriptionTray cooling module
Temperature Setting6-40°C
Unit SizeEach
Catalog Number
Unit Size
Each
Description
With tray cooling
Price (USD)
Full specifications
DescriptionWith tray cooling
IncludesDetector: In-line flow-through UV spectrophotometer
Capacity

Label Free: 36 samples per run

TMT11: 33 samples per run

TMTPro: 32 samples per run

Temperature Setting6-40°C
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Without tray cooling
Price (USD)
Full specifications
DescriptionWithout tray cooling
IncludesDetector: In-line flow-through UV spectrophotometer
Capacity

Label Free: 36 samples per run

TMT11: 33 samples per run

TMTPro: 32 samples per run

Unit SizeEach
Catalog Number
Unit Size
Each
Description
Replacement needle
Price (USD)
Full specifications
DescriptionReplacement needle
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Replacement 1 mL dilutor syringe
Price (USD)
Full specifications
DescriptionReplacement 1 mL dilutor syringe
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Periodic Maintenance kit, end user
Price (USD)
Full specifications
DescriptionPeriodic Maintenance kit, end user
Unit SizeEach
Showing 6 of 6
Catalog NumberSpecificationsUnit SizeDescriptionPrice (USD)
C0960-01-00111Full specifications
EachTray cooling module Request A Quote
DescriptionTray cooling module
Temperature Setting6-40°C
Unit SizeEach
C0960-01-00109Full specifications
EachWith tray coolingRequest A Quote
DescriptionWith tray cooling
IncludesDetector: In-line flow-through UV spectrophotometer
Capacity

Label Free: 36 samples per run

TMT11: 33 samples per run

TMTPro: 32 samples per run

Temperature Setting6-40°C
Unit SizeEach
C0960-01-00110Full specifications
EachWithout tray coolingRequest A Quote
DescriptionWithout tray cooling
IncludesDetector: In-line flow-through UV spectrophotometer
Capacity

Label Free: 36 samples per run

TMT11: 33 samples per run

TMTPro: 32 samples per run

Unit SizeEach
C0950-01-01219Full specifications
EachReplacement needleRequest A Quote
DescriptionReplacement needle
Unit SizeEach
1R77010-1136Full specifications
EachReplacement 1 mL dilutor syringeRequest A Quote
DescriptionReplacement 1 mL dilutor syringe
Unit SizeEach
C0950-01-0013Full specifications
EachPeriodic Maintenance kit, end userRequest A Quote
DescriptionPeriodic Maintenance kit, end user
Unit SizeEach
Showing 6 of 6

The AccelerOme automated sample preparation platform features include:

  • Liquid Handling Robot with a capacity to process up to 36 label-free samples, 33 Thermo Scientific™ TMT11plex isobaric labeling reagent samples or 32 Thermo Scientific™ TMTpro™ 16plex label reagent samples per session
  • Processes sample types consisting of cells, biofluids, tissues, and purified proteins
  • Capable of processing sample amounts ranging from 10 to 100 μg
  • Performs in-line SPE clean-up
  • Measures the final peptide concentration per sample using an in-line spectrophotometer


Operational ease of use:

  • Onscreen graphical wizard to guide users through experimental setup and operation
  • Experiment designer software provides estimates of statistical power for planned experiments
  • Method set-up page to create study/sample name, factor, well location in-plate format and injection volume that can be exported in an acquisition queue to Orbitrap mass spectrometers

Commercially available kits:

  • Thermo Scientific™ reagent kits are pre-made and ready to use in plate format
  • Commercially manufactured kits ensuring product consistency
  • Kits available for label-free and TMT multiplexing in different scaling formats

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text