One Shot™ PIR2 Chemically Competent E. coli
One Shot&trade; PIR2 Chemically Competent <i>E. coli</i>
Invitrogen™

One Shot™ PIR2 Chemically Competent E. coli

One Shot PIR2 Chemically Competent E. coli cells are for use with vectors that contain the R6Kγ origin of replicationRead more
Promo ImagePromotion
Promo code:P5906365
Don't miss out: buy 3, only pay for 2
Promo ImagePromotion
Promo code:RPUZZ25
Stock up on essentials to piece your discovery together
Have Questions?
Catalog NumberQuantity
C111110Promo Image11 x 50 μL/tube
Catalog number C111110
Price (USD)
358.00
Special offer
Online exclusive
392.00
Save 34.00 (9%)
Each
In stock
Add to cart
Quantity:
11 x 50 μL/tube
Recurring order eligible. Learn more »
Price (USD)
358.00
Special offer
Online exclusive
392.00
Save 34.00 (9%)
Each
Add to cart
Ask our AI about this Product

One Shot PIR2 Chemically Competent E. coli cells are for use with vectors that contain the R6Kγ origin of replication (e.g., pUni/V5-His-TOPO for recombination with an Echo cloning system). The pir gene encodes the replication protein π, which is required to replicate and maintain plasmids containing the R6Kγ origin. The PIR2 strain contains the wild-type pir gene that maintains the donor at ∼15 copies per cell. This strain is recommended for maintaining constructs that express toxic genes or for libraries. PIR2 is made chemically competent and can achieve high transformation efficiency of >1 x 108 cfu/μg supercoiled plasmid.

R6Kγ allows tight control and selection and has been used in various applications including genome engineering, mutagenesis studies, bacterial artificial chromosome (BAC) construction, high-capacity extrachromosomal vector systems, and suicide plasmid construction.

One Shot PIR2 Chemically Competent E. coli cells offer:
• Wild-type pir gene to maintain vectors that contain the R6Kγ ori at ∼15 copies per cell
• Transformation efficiencies of >1 x 108 cfu/μg
endA1 mutation that enables increased plasmid DNA yield and quantity
recA1 mutation that reduces the occurrence of unwanted recombination in cloned DNA
hsdR for efficient transformation of unmethylated DNA from PCR applications

Convenient and efficient format
PIR2 Chemically Competent E. coli cells are supplied in the convenient, single-reaction One Shot format. Each tube contains enough cells for one transformation, reducing freeze-thaw cycles and money wasted on unused cells.

Genotype
F∆lac169 rpoS(am) robA1 creC510 hsdR514 endA recA1 uidA(∆MluI)::pir
Genetic marker descriptions

Find the strain and format that fit your needs
Multiple DH strains are available in chemically competent and electrocompetent cell formats.
We offer the PIR1 strain that carries mutated pir gene and maintain vectors at ∼250 copies per cell.
MultiShot formats are available for high throughput applications.
Explore bacterial growth media formats.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Antibiotic Resistance BacterialNo
Blue/White ScreeningNo
Cloning Methylated DNANo
Contains F' EpisomeNo
High-throughput CompatibilityLow
Improves Plasmid QualityYes (endA1)
Preparing Unmethylated DNANo
Product LineOne Shot™
Product TypeChemically Competent Cells
Quantity11 x 50 μL/tube
Reduces RecombinationYes (recA1)
Shipping ConditionDry Ice
T1 Phage - Resistant (tonA)No
FormatTube
SpeciesE. coli
Unit SizeEach
Contents & Storage
• One Shot PIR2 Chemically Competent E. coli (11 x 50 μL)
Store Competent Cells at –80°C.

• pUC19 plasmid (50 μL at 10 pg/μL)
Store pUC19 plasmid at –20°C.

• S.O.C. Medium
Store S.O.C. Medium at 4°C or room temperature.

Ready-to-use Bacterial Growth Media

Ready-to-use Bacterial Growth Media

See how these mediums can help with critical aspects of cloning!

Gibco LB Broth

Invitrogen S.O.C. Medium

Invitrogen One Shot LB Agar*

*Only available in North America and selected European countries

Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Figures

Documents & Downloads

Certificates

0 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection