One Shot™ TOP10 Chemically Competent E. coli
One Shot&trade; TOP10 Chemically Competent <i>E. coli</i>
Invitrogen™

One Shot™ TOP10 Chemically Competent E. coli

One Shot TOP10 Chemically Competent E. coli are ideal for high-efficiency cloning and plasmid DNA propagation and are provided atRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
C40401011 x 50 μL
C40400321 x 50 μL
C40400642 x 50 μL
Catalog number C404010
Price (EUR)
279,65
Online Exclusive
340,00
Save 60,35 (18%)
Each
-
Add to cart
Quantity:
11 x 50 μL
Price (EUR)
279,65
Online Exclusive
340,00
Save 60,35 (18%)
Each
Add to cart
Ask our AI about this Product

One Shot TOP10 Chemically Competent E. coli are ideal for high-efficiency cloning and plasmid DNA propagation and are provided at a transformation efficiency of 1 x 109 cfu/μg plasmid DNA. These cells allow stable replication of high-copy number plasmids and are the same competent cells that come with many of our cloning kits. TOP10 E. coli cells are genetically similar to the DH10B strain and have been reported to be more resilient to stress conditions like osmotic shock and acidic pH stress.

One Shot TOP10 cells:
• Maximize cloning efficiency in a single-tube format
• Provide enhanced genomic DNA cloning capabilities

TOP10 Chemically Competent E. coli cells carry mutations in the methylation-dependent restriction system (mcrA, mcrBC, and mrr) allowing the cloning of both prokaryotic and eukaryotic genomic DNA, as well as efficient plasmid rescue from eukaryotic genomes. Similar to other DH strains, TOP10 has the lacZΔM15 genotype, providing for the option of blue-white screening on plates containing either X-Gal or Bluo-Gal. The inclusion of recA1 and endA1 mutations increase insert stability and improve the quality of plasmid DNA prepared from minipreps.

One Shot TOP10 Chemically Competent E. coli cells offer:
• Transformation efficiencies of >1 x 109 cfu/μg
hsdR for efficient transformation of unmethylated DNA from PCR amplifications
mcrA for efficient transformation of methylated DNA from genomic preparations
lacZΔM15 for blue/white color screening of recombinant clones
endA1 for cleaner DNA preparations and better results in downstream applications due to elimination of nonspecific digestion by Endonuclease I
recA1 for reduced occurrence of nonspecific recombination in cloned DNA
• Expression from the lac promoter without IPTG

Easy-to-use One Shot format
TOP10 Chemically Competent E. coli cells are supplied in the convenient, single-reaction One Shot format. The single-tube, single-use format accomodates allows all steps of the transformation protocol, up to plating, to take place in the same tube, helping save time and prevent contamination.

Genotype
FmcrA Δ(mrr-hsdRMS-mcrBC) φ80lacZΔM15 ΔlacX74 recA1 araD139 Δ(ara-leu)7697 galU galK λ–rpsL(StrR) endA1 nupG

Find the strain and format that fit your needs 
We offer other DH strains in chemically competent and electrocompetent cell formats.
The TOP10 strain is available in several MultiShot formats for high throughput applications.
Explore bacterial growth media formats.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Antibiotic Resistance BacterialYes (Streptomycin)
Blue/White ScreeningYes (lacZΔM15)
Cloning Methylated DNAYes (mcrA)
Cloning Unstable DNANo
Contains F' EpisomeNo
High-throughput CompatibilityLow
Improves Plasmid QualityYes (endA1)
PlasmidHigh Copy Plasmid
Preparing Unmethylated DNANo
Product LineOne Shot™
Product TypeChemically Competent Cells
Quantity11 x 50 μL
Reduces RecombinationYes (recA1)
Shipping ConditionDry Ice
T1 Phage - Resistant (tonA)No
Transformation Efficiency LevelHigh Efficiency (>1 x 109 cfu/μg)
FormatTube
SpeciesE. coli (K12)
Unit SizeEach
Contents & Storage
• One Shot TOP10 Chemically Competent E. coli (11 x 50 μL)
Store Competent Cells at –80°C.

• pUC19 DNA (50 μL at 10 pg/μL)
Store pUC19 DNA at –20°C.

• S.O.C. medium (6 mL)
Store S.O.C. Medium at 4°C or room temperature.

Ready-to-use Bacterial Growth Media

Ready-to-use Bacterial Growth Media

See how these mediums can help with critical aspects of cloning!

Gibco LB Broth

Invitrogen S.O.C. Medium

Invitrogen One Shot LB Agar*

*Only available in North America and selected European countries

Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link