Search by lot number or partial lot number
For Use With (Application) | Immunocytochemistry, Immunohistochemistry, Immunofluorescence |
Quantity | 10 mg |
Source | Canavlia ensiformis |
Conjugate | Alexa Fluor Plus 405 |
Cross Reactivity | α-man, α-glc |
Species | C. ensiformis |
Recombinant | Native |
Protein Tag | None |
Excitation/Emission | 405/450 nm |
Label Type | Classic Dyes |
Shipping Condition | Room Temperature |
Protein Family | Lectins |
Form | Lyophilized |
Protein Subtype | Concanavalin A (Con A) |
Research Category | Imaging |
Color | Violet |
Product Line | Alexa Fluor™ |
Purity or Quality Grade | Imaging Quality |
Ligand Type | Lectin |
Unit Size | 5 mg |
For Use With (Application) | Immunocytochemistry, Immunohistochemistry, Immunofluorescence |
Quantity | 5 mg |
Source | Canavlia ensiformis |
Conjugate | Alexa Fluor 350 |
Cross Reactivity | α-man, α-glc |
Species | C. ensiformis |
Recombinant | Native |
Protein Tag | None |
Excitation/Emission | 346/442 nm |
Label Type | Alexa Fluor Dyes |
Shipping Condition | Room Temperature |
Expression System | Natural |
Protein Family | Lectins |
Form | Lyophilized |
Protein Subtype | Concanavalin A (Con A) |
Research Category | Imaging |
Color | Blue |
Product Line | Alexa Fluor™ |
Purity or Quality Grade | Imaging Quality |
Ligand Type | Lectin |
Unit Size | 5 mg |
For Use With (Application) | Immunocytochemistry, Immunohistochemistry, Immunofluorescence |
Molecular Weight (g/mol) | 104,000 Dalton |
Quantity | 5 mg |
Source | Canavlia ensiformis |
Regulatory Status | RUO |
Conjugate | Alexa Fluor 488 |
Cross Reactivity | α-man, α-glc |
Species | C. ensiformis |
Recombinant | Native |
Protein Tag | None |
Excitation/Emission | 495/519 nm |
Label Type | Alexa Fluor Dyes |
Shipping Condition | Room Temperature |
Expression System | Natural |
Protein Family | Lectins |
Form | Lyophilized |
Protein Subtype | Concanavalin A (Con A) |
Research Category | Imaging |
Color | Green |
Product Line | Alexa Fluor™ |
Purity or Quality Grade | Imaging Quality |
Ligand Type | Lectin |
Unit Size | 5 mg |
For Use With (Application) | Immunocytochemistry, Immunohistochemistry, Immunofluorescence |
Quantity | 5 mg |
Source | Canavlia ensiformis |
Conjugate | Alexa Fluor 488 |
Cross Reactivity | α-man, α-glc |
Species | C. ensiformis |
Recombinant | Native |
Protein Tag | None |
Excitation/Emission | 495/519 nm |
Label Type | Alexa Fluor Dyes |
Shipping Condition | Room Temperature |
Expression System | Natural |
Protein Family | Lectins |
Form | Lyophilized |
Protein Subtype | Concanavalin A, succinylated |
Research Category | Imaging |
Color | Green |
Product Line | Alexa Fluor™ |
Purity or Quality Grade | Imaging Quality |
Ligand Type | Lectin |
Unit Size | 5 mg |
For Use With (Application) | Immunocytochemistry, Immunohistochemistry, Immunofluorescence |
Quantity | 5 mg |
Source | Canavlia ensiformis |
Conjugate | Alexa Fluor 594 |
Cross Reactivity | α-man, α-glc |
Species | C. ensiformis |
Recombinant | Native |
Protein Tag | None |
Excitation/Emission | 590/617 nm |
Label Type | Alexa Fluor Dyes |
Shipping Condition | Room Temperature |
Expression System | Natural |
Protein Family | Lectins |
Form | Lyophilized |
Protein Subtype | Concanavalin A (Con A) |
Research Category | Imaging |
Color | Red |
Product Line | Alexa Fluor™ |
Purity or Quality Grade | Imaging Quality |
Ligand Type | Lectin |
Unit Size | 5 mg |
For Use With (Application) | Immunocytochemistry, Immunohistochemistry, Immunofluorescence |
Quantity | 5 mg |
Source | Canavlia ensiformis |
Conjugate | Alexa Fluor 633 |
Cross Reactivity | α-man, α-glc |
Species | C. ensiformis |
Recombinant | Native |
Protein Tag | None |
Excitation/Emission | 632/647 nm |
Label Type | Alexa Fluor Dyes |
Shipping Condition | Room Temperature |
Expression System | Natural |
Protein Family | Lectins |
Form | Lyophilized |
Protein Subtype | Concanavalin A (Con A) |
Research Category | Imaging |
Color | Deep Red |
Product Line | Alexa Fluor™ |
Purity or Quality Grade | Imaging Quality |
Ligand Type | Lectin |
Unit Size | 5 mg |
For Use With (Application) | Immunocytochemistry, Immunohistochemistry, Immunofluorescence |
Quantity | 5 mg |
Source | Canavlia ensiformis |
Conjugate | Alexa Fluor 647 |
Cross Reactivity | α-man, α-glc |
Species | C. ensiformis |
Recombinant | Native |
Protein Tag | None |
Excitation/Emission | 650/668 nm |
Label Type | Alexa Fluor Dyes |
Shipping Condition | Room Temperature |
Expression System | Natural |
Protein Family | Lectins |
Form | Lyophilized |
Protein Subtype | Concanavalin A (Con A) |
Research Category | Imaging |
Color | Deep Red |
Product Line | Alexa Fluor™ |
Purity or Quality Grade | Imaging Quality |
Ligand Type | Lectin |
Unit Size | 5 mg |
For Use With (Application) | Immunocytochemistry, Immunohistochemistry, Immunofluorescence |
Quantity | 10 mg |
Source | Canavlia ensiformis |
Conjugate | Alexa Fluor Plus 750 |
Cross Reactivity | α-man, α-glc |
Species | C. ensiformis |
Recombinant | Native |
Protein Tag | None |
Excitation/Emission | 750/777 nm |
Label Type | Classic Dyes |
Shipping Condition | Room Temperature |
Protein Family | Lectins |
Form | Lyophilized |
Protein Subtype | Concanavalin A (Con A) |
Research Category | Imaging |
Color | Near-infrared |
Product Line | Alexa Fluor™ |
Purity or Quality Grade | Imaging Quality |
Ligand Type | Lectin |
Unit Size | 1 mg x 5 vials |
For Use With (Application) | Immunocytochemistry, Immunohistochemistry, Immunofluorescence |
Quantity | 10 mg |
Source | Canavlia ensiformis |
Conjugate | Texas Red |
Cross Reactivity | α-man, α-glc |
Species | C. ensiformis |
Recombinant | Native |
Protein Tag | None |
Excitation/Emission | 595/615 nm |
Label Type | Classic Dyes |
Shipping Condition | Room Temperature |
Expression System | Natural |
Protein Family | Lectins |
Form | Lyophilized |
Protein Subtype | Concanavalin A (Con A) |
Research Category | Imaging |
Color | Red |
Product Line | Texas Red™ |
Purity or Quality Grade | Imaging Quality |
Ligand Type | Lectin |
Unit Size | 10 mg |
For Use With (Application) | Immunocytochemistry, Immunohistochemistry, Immunofluorescence |
Quantity | 10 mg |
Source | Canavlia ensiformis |
Conjugate | Tetramethylrhodamine |
Cross Reactivity | α-man, α-glc |
Species | C. ensiformis |
Recombinant | Native |
Protein Tag | None |
Excitation/Emission | 555/580 nm |
Label Type | Classic Dyes |
Shipping Condition | Room Temperature |
Expression System | Natural |
Protein Family | Lectins |
Form | Lyophilized |
Protein Subtype | Concanavalin A (Con A) |
Research Category | Imaging |
Color | Orange |
Purity or Quality Grade | Imaging Quality |
Ligand Type | Lectin |
Unit Size | 10 mg |
Catalog Number | Specifications | Unit Size | Protein Subtype | Conjugate | Color | Excitation/Emission | Price (USD) | Availability | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
C56126 | 5 mg | Concanavalin A (Con A) | Alexa Fluor Plus 405 | Violet | 405/450 nm | 209.65 Online price 241.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
C11254 | 5 mg | Concanavalin A (Con A) | Alexa Fluor 350 | Blue | 346/442 nm | 201.65 Online price 231.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
C11252 | 5 mg | Concanavalin A (Con A) | Alexa Fluor 488 | Green | 495/519 nm | 201.65 Online price 233.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
C21401 | 5 mg | Concanavalin A, succinylated | Alexa Fluor 488 | Green | 495/519 nm | 196.65 Online price 226.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
C11253 | 5 mg | Concanavalin A (Con A) | Alexa Fluor 594 | Red | 590/617 nm | 203.65 Online price 234.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
C21402 | 5 mg | Concanavalin A (Con A) | Alexa Fluor 633 | Deep Red | 632/647 nm | 198.65 Online price 228.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
C21421 | 5 mg | Concanavalin A (Con A) | Alexa Fluor 647 | Deep Red | 650/668 nm | 238.65 Online price 241.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
C56127 | 1 mg x 5 vials | Concanavalin A (Con A) | Alexa Fluor Plus 750 | Near-infrared | 750/777 nm | 209.65 Online price 241.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
C825 | 10 mg | Concanavalin A (Con A) | Texas Red | Red | 595/615 nm | 196.65 Online price 226.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
C860 | 10 mg | Concanavalin A (Con A) | Tetramethylrhodamine | Orange | 555/580 nm | 202.65 Online price 233.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|