Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Catalog Number | Conjugate | Color | Excitation/Emission | Protein Subtype |
---|---|---|---|---|
C11252 | Alexa Fluor 488 | Green | 495/519 nm | Concanavalin A (Con A) |
C11254 | Alexa Fluor 350 | Blue | 346/442 nm | Concanavalin A (Con A) |
C21401 | Alexa Fluor 488 | Green | 495/519 nm | Concanavalin A, succinylated |
C11253 | Alexa Fluor 594 | Red | 590/617 nm | Concanavalin A (Con A) |
C21402 | Alexa Fluor 633 | Deep Red | 632/647 nm | Concanavalin A (Con A) |
C21421 | Alexa Fluor 647 | Deep Red | 650/668 nm | Concanavalin A (Con A) |
C56126 | Alexa Fluor Plus 405 | Violet | 405/450 nm | Concanavalin A (Con A) |
C56127 | Alexa Fluor Plus 750 | Near-infrared | 750/777 nm | Concanavalin A (Con A) |
C825 | Texas Red | Red | 595/615 nm | Concanavalin A (Con A) |
C860 | Tetramethylrhodamine | Orange | 555/580 nm | Concanavalin A (Con A) |
Share catalog number, name or link