Invitrogen™

CellTracker™ CM-DiI Dye

Catalog number: C7001
Have Questions?
Invitrogen™

CellTracker™ CM-DiI Dye

Catalog number: C7001
Have Questions?
Catalog Number
C7001
Unit Size
1 mg
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
20 x 50 µg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
C70011 mg
-
C700020 x 50 µg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
CellTracker CM-DiI is a fluorescent dye well suited for monitoring cell movement or location. This dye is well retained, allowing for multigenerational tracking of cellular movements. And the red excitation/emission spectra are ideal for multiplexing with green fluorescent dyes and proteins.

Need a different emission spectrum or longer tracking? View our other mammalian cell tracking products.

• Easy to use—remove medium, add dye, incubate 30 minutes, and image cells
• Fluorescent signal retention of >72 hours (typically three to six generations)
• Red excitation/emission spectra (553/570 nm maxima) ideal for multiplexing
• Low cytotoxicity—does not affect viability or proliferation

CellTracker CM-DiI fluorescent dye has been designed to freely pass through cell membranes into cells, where it is transformed into cell membrane-impermeant reaction products. CellTracker CM-DiI dye is retained in living cells through several generations. The dye is transferred to daughter cells, but not adjacent cells in a population. CellTracker CM-DiI dye is designed to display fluorescence for at least 72 hours, and the dye exhibits ideal tracking properties: it is stable, nontoxic at working concentrations, well retained in cells, and brightly fluorescent at physiological pH. Additionally, the excitation and emission spectra of CellTracker CM-DiI dye are well separated from GFP (green fluorescent protein) spectra allowing for multiplexing.

Visualize staining your cell without wasting your reagents, antibodies, or time with our new Stain-iT Cell Staining Simulator.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Color
Yellow
For Use With (Equipment)
Fluorescence Microscope
Quantity
1mg
Detection Method
Fluorescent
Product Line
CellTracker™
Shipping Condition
Room Temperature
Label Type
Fluorescent Dye
Product Type
Dye
SubCellular Localization
Cell Membranes, Lipids

Contents & Storage

Store in freezer -5°C to -30°C and protect from light.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text