Cell Culture Contamination Detection Kit
Cell Culture Contamination Detection Kit
Invitrogen™

Cell Culture Contamination Detection Kit

The Cell Culture Contamination Detection Kit is a simple and effective procedure for screening tissue cultures for contamination by microorganisms.Read more
Have Questions?
Catalog NumberQuantity
C7028200 Assays
Catalog number C7028
Price (USD)
407.65
Online Exclusive
419.00
Save 11.35 (3%)
Each
In stock
Add to cart
Quantity:
200 Assays
Price (USD)
407.65
Online Exclusive
419.00
Save 11.35 (3%)
Each
Add to cart
Ask our AI about this Product
The Cell Culture Contamination Detection Kit is a simple and effective procedure for screening tissue cultures for contamination by microorganisms. The kit not only detects contaminants, but it also identifies the contaminant type. Three fluorescent dyes distinctly stain yeast (and other fungi), gram-positive and gram negative bacteria by slide preparation.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodFluorescence
For Use WithFluorescence Microscope
Quantity200 Assays
Shipping ConditionRoom Temperature
FormLiquid
Product TypeCell Culture Contamination Detection
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -30°C) and protect from light.
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Figures

Kunden, die diesen Artikel ansahen, interessierten sich auch für



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
2975913Certificate of AnalysisAug 22, 2024C7028
2820935Certificate of AnalysisJan 24, 2024C7028
2761153Certificate of AnalysisSep 29, 2023C7028
2529640Certificate of AnalysisOct 04, 2022C7028
2529639Certificate of AnalysisAug 12, 2022C7028
5 results displayed, search above for a specific certificate

Safety Data Sheets

Frequently asked questions (FAQs)

This kit can screen tissue cultures for contamination by microorganisms. The kit not only serves to detect the contaminants, it also identifies the contaminant type. Three fluorescent dyes distinctly stain yeast (and other fungi), gram-positive and gram-negative bacteria in slide preparations. The kit provides results within one hour, and, by staining the microorganisms with different fluorescent colors, it helps a scientist choose an appropriate course of action.

Yes, the Cell Culture Contamination Detection Kit will detect/stain mycoplasma. The SYTO-9 cell-permeant dye will stain every cell, including the mammalian cells (if present). Mycoplasma will be seen as small contaminating cells in the medium. It may be challenging to identify Mycoplasma from other possible contaminating species, if present.

We also offer the MycoFluor Mycoplasma Detection Kit (Cat. No. M7006) for mycoplasma detection in lab cultures.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Citations & References (7)

Citations & References
Abstract
Occupancy maps of 208 chromatin-associated proteins in one human cell type.
Authors:
Journal:Nature
PubMed ID:32728244
Cell survival and differentiation with nanocrystalline glass-like carbon using substantia nigra dopaminergic cells derived from transgenic mouse embryos.
Authors:
Journal:PLoS One
PubMed ID:28334019
Supported Cell Membrane Sheets to Monitor Protein Assembly.
Authors:
Journal:Bio Protoc
PubMed ID:33654865
PHF13 is a molecular reader and transcriptional co-regulator of H3K4me2/3.
Authors:
Journal:Elife
PubMed ID:27223324
Elastin is a key factor of tumor development in colorectal cancer.
Authors:
Journal:BMC Cancer
PubMed ID:32171282
7 total citations

Share catalog number, name or link

1x1 image pixel for data collection