Our DQ gelatin is a fluorogenic substrate that can be used to detect protease activity in vitro with high sensitivity. This substrate, formerly available only in our popular EnzChek Gelatinase/Collagenase Assay Kit (E-12055), consists of highly quenched, fluorescein-labeled gelatin. Upon proteolytic digestion, its bright, green fluorescence is revealed and can be used to measure enzymatic activity. The fluorescence increase can be monitored with a fluorescence microplate reader or fluorometer.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection Method
Fluorescence
Form
Powder
Product Line
DQ™
Quantity
5 x 1 mg
Substrate
Protease Substrate
Substrate Properties
Protein-Based Substrate
Target Enzyme
Metalloproteinase
Shipping Condition
Room Temperature
Contents & Storage
Store in freezer (-5 to -30°C) and protect from light.