Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Certifications/Compliance | CE, CSA |
Conductivity | 0.055μS/cm |
Bacterial Content | <1 CFU/mL |
Feed Water Monitoring | No |
Feedwater Source | Tap or Pretreated |
Flow Rate | Up to 2.5 L/min. |
Product Water Type | Type 1 |
Remote Dispenser | Optional |
Resistivity | 18.2 Mohm.cm at 25°C |
TOC Monitor with UV Intensity Monitoring | No |
Total Organic Carbon | <10 ppb |
Type | Water Purification System |
Feedwater Pressure | 100 psig (6.9 bar) |
Modules | 4 |
Particles >0.2µM/mL | <1 μM/mL |
Dimensions (L x W x H) | 7.5 x 37.5 x 28 in. (19 x 95.3 x 71.1 cm) |
Length (English) | 7.5 in. |
Length (Metric) | 19 cm |
Width (English) | 37.5 in. |
Width (Metric) | 95.3 cm |
Height (English) | 28 in. |
Height (Metric) | 71.1 cm |
Voltage | 120 V |
Unit Size | Each |
Certifications/Compliance | CE, CSA |
Conductivity | Type 1: 0.055μS/cm, Type 2: 0.067 to 0.1μS/cm |
Bacterial Content | <1 CFU/mL |
Feed Water Monitoring | No |
Feedwater Source | Tap or Pretreated |
Flow Rate | Up to 2.5 L/min. |
Product Water Type | Type 1 |
Remote Dispenser | Optional |
Resistivity | 18.2 Mohm.cm at 25°C |
TOC Monitor with UV Intensity Monitoring | No |
Total Organic Carbon | <10 ppb |
Type | Water Purification System |
Feedwater Pressure | 100 psig (6.9 bar) |
Modules | 3 |
Particles >0.2µM/mL | <1 μM/mL |
Dimensions (L x W x H) | 7.5 x 31.5 x 28 in. (19 x 80.0 x 71.1 cm) |
Length (English) | 7.5 in. |
Length (Metric) | 19 cm |
Width (English) | 31.5 in. |
Width (Metric) | 80.0 cm |
Height (English) | 28 in. |
Height (Metric) | 71.1 cm |
Voltage | 120 V |
Unit Size | Each |
Certifications/Compliance | CE, CSA |
Conductivity | 0.055μS/cm |
Bacterial Content | <1 CFU/mL |
Feed Water Monitoring | No |
Feedwater Source | Tap or Pretreated |
Flow Rate | Up to 2.5 L/min. |
Product Water Type | Type 1 |
Remote Dispenser | Optional |
Resistivity | 18.2 Mohm.cm at 25°C |
TOC Monitor with UV Intensity Monitoring | No |
Total Organic Carbon | <10 ppb |
Type | Water Purification System |
Feedwater Pressure | 100 psig (6.9 bar) |
Modules | 3 |
Particles >0.2µM/mL | <1 μM/mL |
Dimensions (L x W x H) | 7.5 x 31.5 x 28 in. (19 x 80.0 x 71.1 cm) |
Length (English) | 7.5 in. |
Length (Metric) | 19 cm |
Width (English) | 31.5 in. |
Width (Metric) | 80.0 cm |
Height (English) | 28 in. |
Height (Metric) | 71.1 cm |
Voltage | 240 V |
Unit Size | Each |
Certifications/Compliance | CE, CSA |
Conductivity | 0.055μS/cm |
Bacterial Content | <1 CFU/mL |
Feed Water Monitoring | No |
Feedwater Source | Tap or Pretreated |
Flow Rate | Up to 2.5 L/min. |
Product Water Type | Type 1 |
Remote Dispenser | Optional |
Resistivity | 18.2 Mohm.cm at 25°C |
TOC Monitor with UV Intensity Monitoring | No |
Total Organic Carbon | <10 ppb |
Type | Water Purification System |
Feedwater Pressure | 100 psig (6.9 bar) |
Modules | 4 |
Particles >0.2µM/mL | <1 μM/mL |
Dimensions (L x W x H) | 7.5 x 37.5 x 28 in. (19 x 95.3 x 71.1 cm) |
Length (English) | 7.5 in. |
Length (Metric) | 19 cm |
Width (English) | 37.5 in. |
Width (Metric) | 95.3 cm |
Height (English) | 28 in. |
Height (Metric) | 71.1 cm |
Voltage | 240 V |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Modules | Voltage | Price (USD) | Availability | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
D4641 | Each | 4 | 120 V | 5,665.00 Online price 6,970.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
D4631 | Each | 3 | 120 V | 5,994.00 Online price 6,660.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
D4632-33 | Each | 3 | 240 V | Request A Quote | - | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
D4642-33 | Each | 4 | 240 V | 6,273.00 Online price 6,970.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Ordering Information:
Optional accessories include Remote Dispenser (D8952), Low Pressure Pump Protector (D2706), and Low Water Level Pump Protector (D0603, D0606). These products, as well as replacement cartridges and consumables, are available separately. For details, contact your sales representative.
Includes:
Wall bracket
Requires:
O-rings included with cartridge pack (sold separately)
Recommended for:
AA, GS, buffer preps, feedwater for analytical instruments or labware washers
Note:
If feeding E-Pure from a storage reservoir, Low Water Pump Protector is recommended to protect pump from damage if reservoir water level falls below system requirements. Order separately (D0603 for 120V, D0606 for 240V).