Informational:Inquire about OEM or Commercial Supply version of this product here.
Thermo Scientific™

PagI (BspHI) (10 U/μL)

Catalog number: ER1282
Have Questions?
Thermo Scientific™

PagI (BspHI) (10 U/μL)

Catalog number: ER1282
Have Questions?
Catalog Number
ER1282
Unit Size
2,000 units
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
400 units
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
ER12822,000 units
-
ER1281400 units
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
5'  T ↓C  A  T  G  A   3' 
3'  A  G  T  A  C ↑T   5' 

Thermo Scientific PagI (BspHI) restriction enzyme recognizes T^CATGA sites and cuts best at 37°C in O buffer (isoschizomers: BspHI, CciI, RcaI). See Reaction Conditions for Restriction Enzymes for a table of enzyme activity, conditions for double digestion, and heat inactivation for this and other restriction enzymes. Note: Also available as a FastDigest enzyme for rapid DNA digestion.

Thermo Scientific conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.

Features

• Superior quality—stringent quality control and industry leading manufacturing process
• Convenient color-coded Five Buffer System
• Includes universal Tango buffer for double-digestions
• BSA premixed in reaction buffers
• Wide selection of restriction endonuclease specificities

Applications

• Molecular cloning
• Restriction site mapping
• Genotyping
• Southern blotting
• Restriction fragment length polymorphism (RFLP)
• SNP

Note: Low salt, high glycerol (> 5%) concentrations, pH > 8.0 or a large excess of enzyme may result in star activity. PagI cleavage is impaired by overlapping dam methylation. To avoid dam methylation, use a dam-, dcm- strain such as GM2163. For methylation sensitivity, refer to product specifications.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Concentration
10 U/μL
Methylation Sensitivity
Dam Methylation-Sensitive, Not CpG Methylation-Sensitive, Not Dcm Methylation-Sensitive
Enzyme
PagI (BspHI)
Compatible Buffer
10X Buffer O
Sensitive to Heat Inactivation
Yes
Optimal Reaction Temperature
37°C
Type IIS RE
No
Quantity
2000 U
Product Type
Restriction Enzyme
Research Category
Traditional Cloning

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text