Informational:Inquire about OEM or Commercial Supply version of this product here.
Thermo Scientific™

BseXI (BbvI) (3 U/μL)

Catalog number: ER1451
Have Questions?
Thermo Scientific™

BseXI (BbvI) (3 U/μL)

Catalog number: ER1451
Have Questions?
Catalog Number
ER1451
Unit Size
100 units
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
500 units
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
ER1451100 units
-
ER1452500 units
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
5'  G  C  A  G  C  N8 3'
3'  C  G  T  C  G  N12 5'

Thermo Scientific BseXI (BbvI) restriction enzyme recognizes GCAGC(8/12)^ sites and cuts best at 65°C in its own unique buffer (isoschizomers: BbvI, BstV1I). See Reaction Conditions for Restriction Enzymes for a table of enzyme activity, conditions for double digestion, and heat inactivation for this and other restriction enzymes. Note: Also available as a FastDigest enzyme for rapid DNA digestion.

Thermo Scientific conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.

Features

• Superior quality—stringent quality control and industry leading manufacturing process
• Convenient color-coded Five Buffer System
• Includes universal Tango buffer for double-digestions
• BSA premixed in reaction buffers
• Wide selection of restriction endonuclease specificities

Applications

• Molecular cloning
• Restriction site mapping
• Genotyping
• Southern blotting
• Restriction fragment length polymorphism (RFLP)
• SNP

Note: Assayed using pBR322 DNA (#SD0041). Incubation at 37°C results in 10% activity. Greater than 40-fold overdigestion with BseXI may result in star activity. BseXI may remain associated with the cleaved DNA. This may cause DNA band shifting during electrophoresis. To avoid atypical DNA band patterns, use the 6X DNA Loading Dye & SDS Solution for sample preparation or heat the digested DNA in the presence of SDS prior to electrophoresis. For methylation sensitivity, refer to product specifications.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Concentration
3 U/μL
Methylation Sensitivity
Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive, Not CpG Methylation-Sensitive
Enzyme
BseXI (BbvI)
Compatible Buffer
Unique Buffer (10X Buffer BseXI (BbvI))
Sensitive to Heat Inactivation
Yes
Optimal Reaction Temperature
65°C
Type IIS RE
Yes
Quantity
100 U
Product Type
Restriction Enzyme
Research Category
Scarless Cloning

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text