Informational:Inquire about OEM or Commercial Supply version of this product here.
Thermo Scientific™

AjuI (5 U/μL)

Catalog number: ER1951
Have Questions?
Thermo Scientific™

AjuI (5 U/μL)

Catalog number: ER1951
Have Questions?
Catalog Number
ER1951
Unit Size
100 units
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
ER1951100 units
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
5'  N7G  A  A  N7T  T  G  G  N11 3'
3'  N12C  T  T  N7A  A  C  C  N6 5'

Thermo Scientific AjuI restriction enzyme recognizes ^(7/12)GAA(N)7TTGG(11/6)^ sites and cuts best at 37°C in R (+SAM) buffer. See Reaction Conditions for Restriction Enzymes for a table of enzyme activity, conditions for double digestion, and heat inactivation for this and other restriction enzymes. Note: Also available as a FastDigest enzyme for rapid DNA digestion.

Thermo Scientific conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.

Features

• Superior quality—stringent quality control and industry leading manufacturing process
• Convenient color-coded Five Buffer System
• Includes universal Tango buffer for double-digestions
• BSA premixed in reaction buffers
• Wide selection of restriction endonuclease specificities

Applications

• Molecular cloning
• Restriction site mapping
• Genotyping
• Southern blotting
• Restriction fragment length polymorphism (RFLP)
• SNP

Note: Complete cleavage of some substrates with AjuI is difficult to achieve. AjuI concentration is determined by the maximum cleavage level achieved when no change in the fragmentation pattern is observed with further enzyme increase. AjuI produces double-strand cuts on both sides of the interrupted recognition site. In certain sequence contexts, the cleavage position may be shifted by one base pair. However, the cleavage position indicated above will predominate.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Concentration
5 U/μL
Methylation Sensitivity
Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive, Not CpG Methylation-Sensitive
Enzyme
Aju I
Compatible Buffer
10X Buffer R
Sensitive to Heat Inactivation
Yes
Optimal Reaction Temperature
37°C
Type IIS RE
Yes
Product Type
Restriction Enzyme
Research Category
Traditional Cloning

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text