Informational:Inquire about OEM or Commercial Supply version of this product here.
Thermo Scientific™

Nt.Bpu10I (5 U/μL)

Catalog number: ER2011
Have Questions?
Thermo Scientific™

Nt.Bpu10I (5 U/μL)

Catalog number: ER2011
Have Questions?
Catalog Number
ER2011
Unit Size
1,000 units
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
ER20111,000 units
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
5'  C  C ↓T  N  A  G  C   3' 
3'  G  G  A  N  T  C  G   5' 

Thermo Scientific Nt.Bpu10I nicking enzyme recognizes CC^TNAGC sites and nicks best at 37°C in R buffer. See Reaction Conditions for Restriction Enzymes for a table of enzyme activity, conditions for double digestion, and heat inactivation for this and other restriction enzymes.

Thermo Scientific conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.

Features

• Superior quality—stringent quality control and industry leading manufacturing process
• Convenient color-coded Five Buffer System
• Includes universal Tango buffer for double-digestions
• BSA premixed in reaction buffers
• Wide selection of restriction endonuclease specificities

Applications
Production of single-stranded circular DNA from supercoiled double-stranded plasmids in vitro with subsequent use in DNA sequencing, site-specific mutagenesis, etc Creation of nested deletions Vector preparation for ligation independent cloning method Preparations of covalently closed, double- stranded linear DNA molecules

Note: Nt.Bpu10I may remain associated with the cleaved DNA. This may cause DNA band shifting during electrophoresis. To avoid atypical DNA band patterns, use the 6X DNA Loading Dye & SDS Solution for sample preparation or heat the digested DNA in the presence of SDS prior to electrophoresis.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Concentration
5 U/μL
Enzyme
Nt.Bpu10I
Compatible Buffer
10X Buffer R
Sensitive to Heat Inactivation
Yes
Optimal Reaction Temperature
37°C
Type IIS RE
No
Product Type
Restriction Enzyme
Research Category
Traditional Cloning

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text