EASY-Spray™ HPLC Columns
EASY-Spray™ HPLC Columns
Thermo Scientific™

EASY-Spray™ HPLC Columns

Robust capillary flow HPLC columns for sensitive protein, peptide and mAbs separation using the stand alone or integrated easy to use design.

Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Diameter (Metric)Particle SizePacking Material
ES906150 mm150 μm2 μmSpherical, Fully Porous, Ultrapure Silica
ES900150 mm75 μm3 μmSpherical, Fully Porous, Ultrapure Silica
ES901150 mm50 μm2 μmSpherical, Fully Porous, Ultrapure Silica
ES902250 mm75 μm2 μmSpherical, Fully Porous, Ultrapure Silica
ES903500 mm75 μm2 μmSpherical, Fully Porous, Ultrapure Silica
ES904150 mm75 μm2 μmSpherical, Fully Porous, Ultrapure Silica
ES905750 mm75 μm2 μmSpherical, Fully Porous, Ultrapure Silica
ES911150 mm75 μm2.6 μmSpherical, Solid Core, Ultrapure Silica
ES907150 mm150 μm4 μmPSDVB (Polystyrene Divinylbenzene)
Catalog number ES906
Price (USD)
1,883.62
Each
Add to cart
Length (Metric):
150 mm
Diameter (Metric):
150 μm
Particle Size:
2 μm
Packing Material:
Spherical, Fully Porous, Ultrapure Silica
Price (USD)
1,883.62
Each
Add to cart
Ask our AI about this Product

Ensure robust nano and capillary flow into your LC-MS using Thermo Scientific™ EASY-Spray™ HPLC Columns. The integrated column/emitter design eliminates dead volume and is temperature-controlled for maximum reliability and performance. Rigorously tested to ensure maximum quality, these columns deliver maximum simplicity and ease-of-use. The capillary flow HPLC columns provide sensitive protein, peptide, and monoclonal antibody (mAb) separation. They give proteomics researchers more than ever before: more throughput, more sensitivity, more separation power and more ease of use.

EASY-Spray ̶ Fully integrated HPLC column/temperature/emitter design

  • Integrated fused-silica emitter delivers an exceptionally stable spray
  • Integrated column with temperature control provides maximum reliability and performance
  • A single nanoViper connection between the HPLC column and the MS ion source eliminates dead volumes
  • Thermo Scientific™ nanoViper™ fingertight fitting prevents overtightening and bad connections

Easy Spray Design Benefits

  • Uncompromised performance and ease-of-use in nano LC-MS
  • Ultra-sharp peaks
  • Outstanding peak capacity
  • Maximized peptide coverage
  • Outstanding column-to-column reproducibility and reliability
  • Outstanding peak capacity for comprehensive proteome characterization
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Column FormatAnalytical Column, EASY-Spray
Column TypeReversed Phase
EndcappedYes
Max. Pressure14500 psi (1000 bar)
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Stationary PhaseC18
Surface Area450 m²/g
Temperature60°C
USP TypeL1
Carbon Load15%
Diameter (Metric)150 μm
Length (Metric)150 mm
Particle Size2 μm
PhaseReversed Phase
Pore Size100 Å
Product LineEasy Spray
pH2 to 8
Unit SizeEach

Low-flow chromatography consumables reference guide

Learn about the portfolio of Thermo Scientific consumables for LC-MS proteomics research and the value they can bring to your lab.

Download now

 

Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection