Informational:Inquire about OEM or Commercial Supply version of this product here.
Thermo Scientific™

Phire Reaction Buffer

Catalog number: F524L
Have Questions?
Thermo Scientific™

Phire Reaction Buffer

Catalog number: F524L
Have Questions?
Catalog Number
F524L
Unit Size
4 x 1.5 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
F524L4 x 1.5 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

The reaction buffer for Thermo Scientific Phire Hot Start II DNA Polymerase is available in three formats: non-colored version with and without detergents (Cat. No. F-524L and F-525L, respectively) or Green version (F-527L) containing a density reagent and two tracking dyes for direct loading of PCR products on gel. The colored buffer does not interfere with enzyme performance and is compatible with downstream applications such as DNA sequencing, ligation and restriction digestion.

The concentration of Mg2+ in the reaction buffers is optimized to work well for most amplicons. The buffer contains 1.5 mM MgCl2 at final reaction concentration. Separate tubes of MgCl2 and DMSO are provided for further optimization.

Related Products

  • Phire Green Reaction Buffer (Cat. No. F527L)
  • Phire Reaction Buffer, detergent-free (F525L)
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Polymerase
Phire Hot Start II DNA Polymerase
Quantity
4 x 1.5 mL
Shipping Condition
Wet or Dry Ice
For Use With (Application)
Standard PCR
Concentration
5X
Volume
6 mL

Contents & Storage

• 5X Phire Reaction Buffer, 4 x 1.5 mL
• 50 mM MgCl2, 2 x 1.5 mL
• DMSO, 0.5 mL

Store at -20°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text