Thermo Scientific™

Phusion Green HF Buffer Pack

Catalog number: F538L
Have Questions?
Thermo Scientific™

Phusion Green HF Buffer Pack

Catalog number: F538L
Have Questions?
Catalog Number
F538L
Unit Size
4 x 1.5 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
F538L4 x 1.5 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

Two optimized buffers are available for Thermo Scientific Phusion High-Fidelity DNA Polymerases:
• 5X Phusion HF Buffer (F-518L, F-538L (Phusion Green), F-520L (Detergent-free))
• 5X Phusion GC Buffer (F-519L, F-539L (Phusion Green), F-521L (Detergent-free))

The error rate of Phusion DNA Polymerase in HF Buffer (4.4 x 10-7) is lower than that in GC Buffer (9.5 x 10-7). Therefore, the HF Buffer should be used as the default buffer for high-fidelity amplification. However, GC Buffer can improve the performance of Phusion DNA Polymerase on some difficult or long templates, such as GC-rich templates or those with complex secondary structures. For applications such as microarray or DHPLC, where the DNA templates need to be free of detergents, we recommend using detergent-free reaction buffers.

5X Phusion Green Buffers include a density reagent and two tracking dyes for direct loading of PCR products on a gel. The colored buffers do not interfere with the enzyme performance and are compatible with downstream applications such as DNA sequencing, ligation and restriction digestion.

Related Products
Phusion HF Buffer Pack
Phusion HF Buffer Pack, detergent-free
Phusion GC Buffer Pack
Phusion GC Buffer Pack, detergent-free
Phusion Green GC Buffer Pack

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Polymerase
Phusion High-Fidelity DNA Polymerase
Shipping Condition
Wet or Dry Ice
For Use With (Application)
Standard PCR, High-fidelity PCR
Starting Material
DNA
Volume
4 x 1.5 mL

Contents & Storage

• 5X Phusion Green HF Reaction Buffer (4 x 1.5 mL)
• 50 mM MgCl2 (4 x 1.5 mL)
• DMSO (0.5 mL)

Store at –20°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text