F6020C-33-80 Full specifications
Each Digital programmable with 4 programs (C1) 240 V CSA approved, CE marked Request A Quote - - Capacity (English) 0.5 cu.ft. Certifications/Compliance CSA approved, CE marked Description F6020C-33-80 Capacity (Metric) 14 L Temperature (Metric) 100°C to 1200°C Type Multiprogram Power Consumption 4400 w Shelves Stainless Steel Temperature Control Digital programmable with 4 programs (C1) Temperature Stability ±1.5 at 1000°C Temperature Uniformity ±4.5 at 1000°C Dimensions (D x W x H) Interior 10 x 12.8 x 6.8 in. (25 x 33 x 18 cm) Dimensions (L x W x H) Exterior 20.1 x 19.1 x 21 in. (51 x 48.5 x 53.3 cm) Length (English) 20.1 in. Length (Metric) 51 cm Width (English) 19.1 in. Width (Metric) 48.5 cm Width (Metric) Interior 33 cm Height (English) 21 in. Height (English) Interior 6.8 in. Height (Metric) 53.3 cm Height (Metric) Interior 18 cm Depth (English) Interior 10 in. Depth (Metric) Interior 25 cm Shipping Weight (English) 134 lb. Shipping Weight (Metric) 60.8 kg Electrical Requirements 240 V 50/60 Hz Voltage 240 V Amperage 18.3 A Frequency 50/60 Hz Unit Size Each
F6010 Full specifications
Each Digital single setpoint (A1) 240 V CSA approved 4,239.00 Online price 4,710.00
- Capacity (English) 0.5 cu.ft. Certifications/Compliance CSA approved Description F6010 Capacity (Metric) 14 L Temperature (Metric) 100°C to 1200°C Type Single Setpoint Power Consumption 3095 w Shelves Stainless Steel Temperature Control Digital single setpoint (A1) Temperature Stability ±0.3 at 1000°C Temperature Uniformity ±2.2 at 1000°C Dimensions (D x W x H) Interior 10 x 12.8 x 6.8 in. (25 x 33 x 18 cm) Dimensions (L x W x H) Exterior 20.1 x 19.1 x 21 in. (51 x 48.5 x 53.3 cm) Length (English) 20.1 in. Length (Metric) 51 cm Width (English) 19.1 in. Width (Metric) 48.5 cm Width (Metric) Interior 33 cm Height (English) 21 in. Height (English) Interior 6.8 in. Height (Metric) 53.3 cm Height (Metric) Interior 18 cm Depth (English) Interior 10 in. Depth (Metric) Interior 25 cm Shipping Weight (English) 134 lb. Shipping Weight (Metric) 60.8 kg Electrical Requirements 240 V 50/60 Hz Voltage 240 V Amperage 12.9 A Frequency 50/60 Hz Unit Size Each
F6018 Full specifications
Each Digital single setpoint (A1) 208 V CSA approved 4,293.00 Online price 4,770.00
- Capacity (English) 0.5 cu.ft. Certifications/Compliance CSA approved Description F6018 Capacity (Metric) 14 L Temperature (Metric) 100°C to 1200°C Type Single Setpoint Power Consumption 2325 w Shelves Stainless Steel Temperature Control Digital single setpoint (A1) Temperature Stability ±0.3 at 1000°C Temperature Uniformity ±2.2 at 1000°C Dimensions (D x W x H) Interior 10 x 12.8 x 6.8 in. (25 x 33 x 18 cm) Dimensions (L x W x H) Exterior 20.1 x 19.1 x 21 in. (51 x 48.5 x 53.3 cm) Length (English) 20.1 in. Length (Metric) 51 cm Width (English) 19.1 in. Width (Metric) 48.5 cm Width (Metric) Interior 33 cm Height (English) 21 in. Height (English) Interior 6.8 in. Height (Metric) 53.3 cm Height (Metric) Interior 18 cm Depth (English) Interior 10 in. Depth (Metric) Interior 25 cm Shipping Weight (English) 134 lb. Shipping Weight (Metric) 60.8 kg Electrical Requirements 208 V 50/60 Hz Voltage 208 V Amperage 11.2 A Frequency 50/60 Hz Unit Size Each
F6020C-33 Full specifications
Each Digital single setpoint with ramp and dwell (B1) 240 V CSA approved, CE marked Request A Quote - - Capacity (English) 0.5 cu.ft. Certifications/Compliance CSA approved, CE marked Description F6020C-33 Capacity (Metric) 14 L Temperature (Metric) 100°C to 1200°C Type Single Setpoint Power Consumption 4400 w Shelves Stainless Steel Temperature Control Digital single setpoint with ramp and dwell (B1) Temperature Stability ±1.5 at 1000°C Temperature Uniformity ±4.5 at 1000°C Dimensions (D x W x H) Interior 10 x 12.8 x 6.8 in. (25 x 33 x 18 cm) Dimensions (L x W x H) Exterior 20.1 x 19.1 x 21 in. (51 x 48.5 x 53.3 cm) Length (English) 20.1 in. Length (Metric) 51 cm Width (English) 19.1 in. Width (Metric) 48.5 cm Width (Metric) Interior 33 cm Height (English) 21 in. Height (English) Interior 6.8 in. Height (Metric) 53.3 cm Height (Metric) Interior 18 cm Depth (English) Interior 10 in. Depth (Metric) Interior 25 cm Shipping Weight (English) 134 lb. Shipping Weight (Metric) 60.8 kg Electrical Requirements 240 V 50/60 Hz Voltage 240 V Amperage 18.3 A Frequency 50/60 Hz Unit Size Each
F6020C-80 Full specifications
Each Digital programmable with 4 programs (C1) 240 V CSA approved Request A Quote - - Capacity (English) 0.5 cu.ft. Certifications/Compliance CSA approved Description F6020C-80 Capacity (Metric) 14 L Temperature (Metric) 100°C to 1200°C Type Multiprogram Power Consumption 4400 w Shelves Stainless Steel Temperature Control Digital programmable with 4 programs (C1) Temperature Stability ±1.5 at 1000°C Temperature Uniformity ±4.5 at 1000°C Dimensions (D x W x H) Interior 10 x 12.8 x 6.8 in. (25 x 33 x 18 cm) Dimensions (L x W x H) Exterior 20.1 x 19.1 x 21 in. (51 x 48.5 x 53.3 cm) Length (English) 20.1 in. Length (Metric) 51 cm Width (English) 19.1 in. Width (Metric) 48.5 cm Width (Metric) Interior 33 cm Height (English) 21 in. Height (English) Interior 6.8 in. Height (Metric) 53.3 cm Height (Metric) Interior 18 cm Depth (English) Interior 10 in. Depth (Metric) Interior 25 cm Shipping Weight (English) 134 lb. Shipping Weight (Metric) 60.8 kg Electrical Requirements 240 V 50/60 Hz Voltage 240 V Amperage 18.3 A Frequency 50/60 Hz Unit Size Each
F6028C Full specifications
Each Digital single setpoint with ramp and dwell (B1) 208 V CSA approved 7,191.00 Online price 7,990.00
- Capacity (English) 0.5 cu.ft. Certifications/Compliance CSA approved Description F6028C Capacity (Metric) 14 L Temperature (Metric) 100°C to 1200°C Type Single Power Consumption 4000 w Shelves Stainless Steel Temperature Control Digital single setpoint with ramp and dwell (B1) Temperature Stability ±1.5 at 1000°C Temperature Uniformity ±4.5 at 1000°C Dimensions (D x W x H) Interior 10 x 12.8 x 6.8 in. (25 x 33 x 18 cm) Dimensions (L x W x H) Exterior 20.1 x 19.1 x 21 in. (51 x 48.5 x 53.3 cm) Length (English) 20.1 in. Length (Metric) 51 cm Width (English) 19.1 in. Width (Metric) 48.5 cm Width (Metric) Interior 33 cm Height (English) 21 in. Height (English) Interior 6.8 in. Height (Metric) 53.3 cm Height (Metric) Interior 18 cm Depth (English) Interior 10 in. Depth (Metric) Interior 25 cm Shipping Weight (English) 134 lb. Shipping Weight (Metric) 60.8 kg Electrical Requirements 208 V 50/60 Hz Voltage 208 V Amperage 19.2 A Frequency 50/60 Hz Unit Size Each
F6028C-80 Full specifications
Each Digital programmable with 4 programs (C1) 208 V CSA approved Request A Quote - - Capacity (English) 0.5 cu.ft. Certifications/Compliance CSA approved Description F6028C-80 Capacity (Metric) 14 L Temperature (Metric) 100°C to 1200°C Type Multiprogram Power Consumption 4000 w Shelves Stainless Steel Temperature Control Digital programmable with 4 programs (C1) Temperature Stability ±1.5 at 1000°C Temperature Uniformity ±4.5 at 1000°C Dimensions (D x W x H) Interior 10 x 12.8 x 6.8 in. (25 x 33 x 18 cm) Dimensions (L x W x H) Exterior 20.1 x 19.1 x 21 in. (51 x 48.5 x 53.3 cm) Length (English) 20.1 in. Length (Metric) 51 cm Width (English) 19.1 in. Width (Metric) 48.5 cm Width (Metric) Interior 33 cm Height (English) 21 in. Height (English) Interior 6.8 in. Height (Metric) 53.3 cm Height (Metric) Interior 18 cm Depth (English) Interior 10 in. Depth (Metric) Interior 25 cm Shipping Weight (English) 134 lb. Shipping Weight (Metric) 60.8 kg Electrical Requirements 208 V 50/60 Hz Voltage 208 V Amperage 19.2 A Frequency 50/60 Hz Unit Size Each
F6030CM Full specifications
Each Digital programmable with 4 programs, 16 segments 240 V CSA approved 9,900.00 Online price 11,000.00
- Capacity (English) 0.5 cu.ft. Certifications/Compliance CSA approved Description F6030CM Capacity (Metric) 14 L Temperature (Metric) 100°C to 1200°C Type Multisegment, Multiprogram Power Consumption 4400 w Shelves Stainless Steel Temperature Control Digital programmable with 4 programs, 16 segments Temperature Stability ±0.2 at 1000°C Temperature Uniformity ±2.2 at 1000°C Dimensions (D x W x H) Interior 10 x 12.8 x 6.8 in. (25 x 33 x 18 cm) Dimensions (L x W x H) Exterior 20.1 x 19.1 x 21 in. (51 x 48.5 x 53.3 cm) Length (English) 20.1 in. Length (Metric) 51 cm Width (English) 19.1 in. Width (Metric) 48.5 cm Width (Metric) Interior 33 cm Height (English) 21 in. Height (English) Interior 6.8 in. Height (Metric) 53.3 cm Height (Metric) Interior 18 cm Depth (English) Interior 10 in. Depth (Metric) Interior 25 cm Shipping Weight (English) 134 lb. Shipping Weight (Metric) 60.8 kg Electrical Requirements 240 V 50/60 Hz Voltage 240 V Amperage 18.3 A Frequency 50/60 Hz Unit Size Each
F6030CM-33 Full specifications
Each Digital programmable with 4 programs, 16 segments 240 V CSA approved, CE marked Request A Quote - - Capacity (English) 0.5 cu.ft. Certifications/Compliance CSA approved, CE marked Description F6030CM-33 Capacity (Metric) 14 L Temperature (Metric) 100°C to 1200°C Type Multisegment, Multiprogram Power Consumption 4400 w Shelves Stainless Steel Temperature Control Digital programmable with 4 programs, 16 segments Temperature Stability ±1.5 at 1000°C Temperature Uniformity ±4.5 at 1000°C Dimensions (D x W x H) Interior 10 x 12.8 x 6.8 in. (25 x 33 x 18 cm) Dimensions (L x W x H) Exterior 20.1 x 19.1 x 21 in. (51 x 48.5 x 53.3 cm) Length (English) 20.1 in. Length (Metric) 51 cm Width (English) 19.1 in. Width (Metric) 48.5 cm Width (Metric) Interior 33 cm Height (English) 21 in. Height (English) Interior 6.8 in. Height (Metric) 53.3 cm Height (Metric) Interior 18 cm Depth (English) Interior 10 in. Depth (Metric) Interior 25 cm Shipping Weight (English) 134 lb. Shipping Weight (Metric) 60.8 kg Electrical Requirements 240 V 50/60 Hz Voltage 240 V Amperage 18.3 A Frequency 50/60 Hz Unit Size Each
F6038CM Full specifications
Each Digital programmable with 4 programs, 16 segments 208 V CSA approved 9,900.00 Online price 11,000.00
- Capacity (English) 0.5 cu.ft. Certifications/Compliance CSA approved Description F6038CM Capacity (Metric) 14 L Temperature (Metric) 100°C to 1200°C Type Multisegment, Multiprogram Power Consumption 4000 w Shelves Stainless Steel Temperature Control Digital programmable with 4 programs, 16 segments Temperature Stability ±1.5 at 1000°C Temperature Uniformity ±4.5 at 1000°C Dimensions (D x W x H) Interior 10 x 12.8 x 6.8 in. (25 x 33 x 18 cm) Dimensions (L x W x H) Exterior 20.1 x 19.1 x 21 in. (51 x 48.5 x 53.3 cm) Length (English) 20.1 in. Length (Metric) 51 cm Width (English) 19.1 in. Width (Metric) 48.5 cm Width (Metric) Interior 33 cm Height (English) 21 in. Height (English) Interior 6.8 in. Height (Metric) 53.3 cm Height (Metric) Interior 18 cm Depth (English) Interior 10 in. Depth (Metric) Interior 25 cm Shipping Weight (English) 134 lb. Shipping Weight (Metric) 60.8 kg Electrical Requirements 208 V 50/60 Hz Voltage 208 V Amperage 19.3 A Frequency 50/60 Hz Unit Size Each