Thermo Scientific™
Thermolyne™ Premium Large Muffle Furnaces
Catalog number: F6020C-80
Have Questions?
Thermo Scientific™
Thermolyne™ Premium Large Muffle Furnaces
Catalog number: F6020C-80
Have Questions?
Conduct industrial applications including ashing organic and inorganic samples and conducting gravimetric analysis with Thermo Scientific™ Thermolyne™ Premium Large Muffle Furnaces. Unit has four heating elements, which are located on the chamber top, bottom and sides for enhanced temperature uniformity. Built-in vent port removes undesirable contaminants and moisture to extend the life of the element and unit.
Catalog Number
F6020C-80
Unit Size
Each
Temperature Control
Digital programmable with 4 programs (C1)
Voltage
240 V
Certifications/Compliance
CSA approved
Price (USD)
Availability
-
Full specifications
Quantity
-
Capacity (English)0.5 cu.ft.
Certifications/ComplianceCSA approved
DescriptionF6020C-80
Capacity (Metric)14 L
Temperature (Metric)100°C to 1200°C
TypeMultiprogram
Power Consumption4400 w
ShelvesStainless Steel
Temperature ControlDigital programmable with 4 programs (C1)
Temperature Stability±1.5 at 1000°C
Temperature Uniformity±4.5 at 1000°C
Dimensions (D x W x H) Interior10 x 12.8 x 6.8 in. (25 x 33 x 18 cm)
Dimensions (L x W x H) Exterior20.1 x 19.1 x 21 in. (51 x 48.5 x 53.3 cm)
Length (English)20.1 in.
Length (Metric)51 cm
Width (English)19.1 in.
Width (Metric)48.5 cm
Width (Metric) Interior33 cm
Height (English)21 in.
Height (English) Interior6.8 in.
Height (Metric)53.3 cm
Height (Metric) Interior18 cm
Depth (English) Interior10 in.
Depth (Metric) Interior25 cm
Shipping Weight (English)134 lb.
Shipping Weight (Metric)60.8 kg
Electrical Requirements240 V 50/60 Hz
Voltage240 V
Amperage18.3 A
Frequency50/60 Hz
Unit SizeEach
Catalog Number
Unit Size
Each
Temperature Control
Digital single setpoint (A1)
Voltage
240 V
Certifications/Compliance
CSA approved
Price (USD)
Availability
-
Full specifications
Quantity
Capacity (English)0.5 cu.ft.
Certifications/ComplianceCSA approved
DescriptionF6010
Capacity (Metric)14 L
Temperature (Metric)100°C to 1200°C
TypeSingle Setpoint
Power Consumption3095 w
ShelvesStainless Steel
Temperature ControlDigital single setpoint (A1)
Temperature Stability±0.3 at 1000°C
Temperature Uniformity±2.2 at 1000°C
Dimensions (D x W x H) Interior10 x 12.8 x 6.8 in. (25 x 33 x 18 cm)
Dimensions (L x W x H) Exterior20.1 x 19.1 x 21 in. (51 x 48.5 x 53.3 cm)
Length (English)20.1 in.
Length (Metric)51 cm
Width (English)19.1 in.
Width (Metric)48.5 cm
Width (Metric) Interior33 cm
Height (English)21 in.
Height (English) Interior6.8 in.
Height (Metric)53.3 cm
Height (Metric) Interior18 cm
Depth (English) Interior10 in.
Depth (Metric) Interior25 cm
Shipping Weight (English)134 lb.
Shipping Weight (Metric)60.8 kg
Electrical Requirements240 V 50/60 Hz
Voltage240 V
Amperage12.9 A
Frequency50/60 Hz
Unit SizeEach
Catalog Number
Unit Size
Each
Temperature Control
Digital single setpoint (A1)
Voltage
208 V
Certifications/Compliance
CSA approved
Price (USD)
Availability
-
Full specifications
Quantity
Capacity (English)0.5 cu.ft.
Certifications/ComplianceCSA approved
DescriptionF6018
Capacity (Metric)14 L
Temperature (Metric)100°C to 1200°C
TypeSingle Setpoint
Power Consumption2325 w
ShelvesStainless Steel
Temperature ControlDigital single setpoint (A1)
Temperature Stability±0.3 at 1000°C
Temperature Uniformity±2.2 at 1000°C
Dimensions (D x W x H) Interior10 x 12.8 x 6.8 in. (25 x 33 x 18 cm)
Dimensions (L x W x H) Exterior20.1 x 19.1 x 21 in. (51 x 48.5 x 53.3 cm)
Length (English)20.1 in.
Length (Metric)51 cm
Width (English)19.1 in.
Width (Metric)48.5 cm
Width (Metric) Interior33 cm
Height (English)21 in.
Height (English) Interior6.8 in.
Height (Metric)53.3 cm
Height (Metric) Interior18 cm
Depth (English) Interior10 in.
Depth (Metric) Interior25 cm
Shipping Weight (English)134 lb.
Shipping Weight (Metric)60.8 kg
Electrical Requirements208 V 50/60 Hz
Voltage208 V
Amperage11.2 A
Frequency50/60 Hz
Unit SizeEach
Catalog Number
Unit Size
Each
Temperature Control
Digital single setpoint with ramp and dwell (B1)
Voltage
240 V
Certifications/Compliance
CSA approved, CE marked
Price (USD)
Availability
-
Full specifications
Quantity
-
Capacity (English)0.5 cu.ft.
Certifications/ComplianceCSA approved, CE marked
DescriptionF6020C-33
Capacity (Metric)14 L
Temperature (Metric)100°C to 1200°C
TypeSingle Setpoint
Power Consumption4400 w
ShelvesStainless Steel
Temperature ControlDigital single setpoint with ramp and dwell (B1)
Temperature Stability±1.5 at 1000°C
Temperature Uniformity±4.5 at 1000°C
Dimensions (D x W x H) Interior10 x 12.8 x 6.8 in. (25 x 33 x 18 cm)
Dimensions (L x W x H) Exterior20.1 x 19.1 x 21 in. (51 x 48.5 x 53.3 cm)
Length (English)20.1 in.
Length (Metric)51 cm
Width (English)19.1 in.
Width (Metric)48.5 cm
Width (Metric) Interior33 cm
Height (English)21 in.
Height (English) Interior6.8 in.
Height (Metric)53.3 cm
Height (Metric) Interior18 cm
Depth (English) Interior10 in.
Depth (Metric) Interior25 cm
Shipping Weight (English)134 lb.
Shipping Weight (Metric)60.8 kg
Electrical Requirements240 V 50/60 Hz
Voltage240 V
Amperage18.3 A
Frequency50/60 Hz
Unit SizeEach
Catalog Number
Unit Size
Each
Temperature Control
Digital programmable with 4 programs (C1)
Voltage
240 V
Certifications/Compliance
CSA approved, CE marked
Price (USD)
Availability
-
Full specifications
Quantity
-
Capacity (English)0.5 cu.ft.
Certifications/ComplianceCSA approved, CE marked
DescriptionF6020C-33-80
Capacity (Metric)14 L
Temperature (Metric)100°C to 1200°C
TypeMultiprogram
Power Consumption4400 w
ShelvesStainless Steel
Temperature ControlDigital programmable with 4 programs (C1)
Temperature Stability±1.5 at 1000°C
Temperature Uniformity±4.5 at 1000°C
Dimensions (D x W x H) Interior10 x 12.8 x 6.8 in. (25 x 33 x 18 cm)
Dimensions (L x W x H) Exterior20.1 x 19.1 x 21 in. (51 x 48.5 x 53.3 cm)
Length (English)20.1 in.
Length (Metric)51 cm
Width (English)19.1 in.
Width (Metric)48.5 cm
Width (Metric) Interior33 cm
Height (English)21 in.
Height (English) Interior6.8 in.
Height (Metric)53.3 cm
Height (Metric) Interior18 cm
Depth (English) Interior10 in.
Depth (Metric) Interior25 cm
Shipping Weight (English)134 lb.
Shipping Weight (Metric)60.8 kg
Electrical Requirements240 V 50/60 Hz
Voltage240 V
Amperage18.3 A
Frequency50/60 Hz
Unit SizeEach
Catalog Number
Unit Size
Each
Temperature Control
Digital single setpoint with ramp and dwell (B1)
Voltage
208 V
Certifications/Compliance
CSA approved
Price (USD)
Availability
-
Full specifications
Quantity
Capacity (English)0.5 cu.ft.
Certifications/ComplianceCSA approved
DescriptionF6028C
Capacity (Metric)14 L
Temperature (Metric)100°C to 1200°C
TypeSingle
Power Consumption4000 w
ShelvesStainless Steel
Temperature ControlDigital single setpoint with ramp and dwell (B1)
Temperature Stability±1.5 at 1000°C
Temperature Uniformity±4.5 at 1000°C
Dimensions (D x W x H) Interior10 x 12.8 x 6.8 in. (25 x 33 x 18 cm)
Dimensions (L x W x H) Exterior20.1 x 19.1 x 21 in. (51 x 48.5 x 53.3 cm)
Length (English)20.1 in.
Length (Metric)51 cm
Width (English)19.1 in.
Width (Metric)48.5 cm
Width (Metric) Interior33 cm
Height (English)21 in.
Height (English) Interior6.8 in.
Height (Metric)53.3 cm
Height (Metric) Interior18 cm
Depth (English) Interior10 in.
Depth (Metric) Interior25 cm
Shipping Weight (English)134 lb.
Shipping Weight (Metric)60.8 kg
Electrical Requirements208 V 50/60 Hz
Voltage208 V
Amperage19.2 A
Frequency50/60 Hz
Unit SizeEach
Catalog Number
Unit Size
Each
Temperature Control
Digital programmable with 4 programs (C1)
Voltage
208 V
Certifications/Compliance
CSA approved
Price (USD)
Availability
-
Full specifications
Quantity
-
Capacity (English)0.5 cu.ft.
Certifications/ComplianceCSA approved
DescriptionF6028C-80
Capacity (Metric)14 L
Temperature (Metric)100°C to 1200°C
TypeMultiprogram
Power Consumption4000 w
ShelvesStainless Steel
Temperature ControlDigital programmable with 4 programs (C1)
Temperature Stability±1.5 at 1000°C
Temperature Uniformity±4.5 at 1000°C
Dimensions (D x W x H) Interior10 x 12.8 x 6.8 in. (25 x 33 x 18 cm)
Dimensions (L x W x H) Exterior20.1 x 19.1 x 21 in. (51 x 48.5 x 53.3 cm)
Length (English)20.1 in.
Length (Metric)51 cm
Width (English)19.1 in.
Width (Metric)48.5 cm
Width (Metric) Interior33 cm
Height (English)21 in.
Height (English) Interior6.8 in.
Height (Metric)53.3 cm
Height (Metric) Interior18 cm
Depth (English) Interior10 in.
Depth (Metric) Interior25 cm
Shipping Weight (English)134 lb.
Shipping Weight (Metric)60.8 kg
Electrical Requirements208 V 50/60 Hz
Voltage208 V
Amperage19.2 A
Frequency50/60 Hz
Unit SizeEach
Catalog Number
Unit Size
Each
Temperature Control
Digital programmable with 4 programs, 16 segments
Voltage
240 V
Certifications/Compliance
CSA approved
Price (USD)
Availability
-
Full specifications
Quantity
Capacity (English)0.5 cu.ft.
Certifications/ComplianceCSA approved
DescriptionF6030CM
Capacity (Metric)14 L
Temperature (Metric)100°C to 1200°C
TypeMultisegment, Multiprogram
Power Consumption4400 w
ShelvesStainless Steel
Temperature ControlDigital programmable with 4 programs, 16 segments
Temperature Stability±0.2 at 1000°C
Temperature Uniformity±2.2 at 1000°C
Dimensions (D x W x H) Interior10 x 12.8 x 6.8 in. (25 x 33 x 18 cm)
Dimensions (L x W x H) Exterior20.1 x 19.1 x 21 in. (51 x 48.5 x 53.3 cm)
Length (English)20.1 in.
Length (Metric)51 cm
Width (English)19.1 in.
Width (Metric)48.5 cm
Width (Metric) Interior33 cm
Height (English)21 in.
Height (English) Interior6.8 in.
Height (Metric)53.3 cm
Height (Metric) Interior18 cm
Depth (English) Interior10 in.
Depth (Metric) Interior25 cm
Shipping Weight (English)134 lb.
Shipping Weight (Metric)60.8 kg
Electrical Requirements240 V 50/60 Hz
Voltage240 V
Amperage18.3 A
Frequency50/60 Hz
Unit SizeEach
Catalog Number
Unit Size
Each
Temperature Control
Digital programmable with 4 programs, 16 segments
Voltage
240 V
Certifications/Compliance
CSA approved, CE marked
Price (USD)
Availability
-
Full specifications
Quantity
-
Capacity (English)0.5 cu.ft.
Certifications/ComplianceCSA approved, CE marked
DescriptionF6030CM-33
Capacity (Metric)14 L
Temperature (Metric)100°C to 1200°C
TypeMultisegment, Multiprogram
Power Consumption4400 w
ShelvesStainless Steel
Temperature ControlDigital programmable with 4 programs, 16 segments
Temperature Stability±1.5 at 1000°C
Temperature Uniformity±4.5 at 1000°C
Dimensions (D x W x H) Interior10 x 12.8 x 6.8 in. (25 x 33 x 18 cm)
Dimensions (L x W x H) Exterior20.1 x 19.1 x 21 in. (51 x 48.5 x 53.3 cm)
Length (English)20.1 in.
Length (Metric)51 cm
Width (English)19.1 in.
Width (Metric)48.5 cm
Width (Metric) Interior33 cm
Height (English)21 in.
Height (English) Interior6.8 in.
Height (Metric)53.3 cm
Height (Metric) Interior18 cm
Depth (English) Interior10 in.
Depth (Metric) Interior25 cm
Shipping Weight (English)134 lb.
Shipping Weight (Metric)60.8 kg
Electrical Requirements240 V 50/60 Hz
Voltage240 V
Amperage18.3 A
Frequency50/60 Hz
Unit SizeEach
Catalog Number
Unit Size
Each
Temperature Control
Digital programmable with 4 programs, 16 segments
Voltage
208 V
Certifications/Compliance
CSA approved
Price (USD)
Availability
-
Full specifications
Quantity
Capacity (English)0.5 cu.ft.
Certifications/ComplianceCSA approved
DescriptionF6038CM
Capacity (Metric)14 L
Temperature (Metric)100°C to 1200°C
TypeMultisegment, Multiprogram
Power Consumption4000 w
ShelvesStainless Steel
Temperature ControlDigital programmable with 4 programs, 16 segments
Temperature Stability±1.5 at 1000°C
Temperature Uniformity±4.5 at 1000°C
Dimensions (D x W x H) Interior10 x 12.8 x 6.8 in. (25 x 33 x 18 cm)
Dimensions (L x W x H) Exterior20.1 x 19.1 x 21 in. (51 x 48.5 x 53.3 cm)
Length (English)20.1 in.
Length (Metric)51 cm
Width (English)19.1 in.
Width (Metric)48.5 cm
Width (Metric) Interior33 cm
Height (English)21 in.
Height (English) Interior6.8 in.
Height (Metric)53.3 cm
Height (Metric) Interior18 cm
Depth (English) Interior10 in.
Depth (Metric) Interior25 cm
Shipping Weight (English)134 lb.
Shipping Weight (Metric)60.8 kg
Electrical Requirements208 V 50/60 Hz
Voltage208 V
Amperage19.3 A
Frequency50/60 Hz
Unit SizeEach
Showing 10 of 10
Catalog NumberSpecificationsUnit SizeTemperature ControlVoltageCertifications/CompliancePrice (USD)AvailabilityQuantity
F6020C-80Full specifications
EachDigital programmable with 4 programs (C1)240 VCSA approvedRequest A Quote--
Capacity (English)0.5 cu.ft.
Certifications/ComplianceCSA approved
DescriptionF6020C-80
Capacity (Metric)14 L
Temperature (Metric)100°C to 1200°C
TypeMultiprogram
Power Consumption4400 w
ShelvesStainless Steel
Temperature ControlDigital programmable with 4 programs (C1)
Temperature Stability±1.5 at 1000°C
Temperature Uniformity±4.5 at 1000°C
Dimensions (D x W x H) Interior10 x 12.8 x 6.8 in. (25 x 33 x 18 cm)
Dimensions (L x W x H) Exterior20.1 x 19.1 x 21 in. (51 x 48.5 x 53.3 cm)
Length (English)20.1 in.
Length (Metric)51 cm
Width (English)19.1 in.
Width (Metric)48.5 cm
Width (Metric) Interior33 cm
Height (English)21 in.
Height (English) Interior6.8 in.
Height (Metric)53.3 cm
Height (Metric) Interior18 cm
Depth (English) Interior10 in.
Depth (Metric) Interior25 cm
Shipping Weight (English)134 lb.
Shipping Weight (Metric)60.8 kg
Electrical Requirements240 V 50/60 Hz
Voltage240 V
Amperage18.3 A
Frequency50/60 Hz
Unit SizeEach
F6010Full specifications
EachDigital single setpoint (A1)240 VCSA approved
-
Capacity (English)0.5 cu.ft.
Certifications/ComplianceCSA approved
DescriptionF6010
Capacity (Metric)14 L
Temperature (Metric)100°C to 1200°C
TypeSingle Setpoint
Power Consumption3095 w
ShelvesStainless Steel
Temperature ControlDigital single setpoint (A1)
Temperature Stability±0.3 at 1000°C
Temperature Uniformity±2.2 at 1000°C
Dimensions (D x W x H) Interior10 x 12.8 x 6.8 in. (25 x 33 x 18 cm)
Dimensions (L x W x H) Exterior20.1 x 19.1 x 21 in. (51 x 48.5 x 53.3 cm)
Length (English)20.1 in.
Length (Metric)51 cm
Width (English)19.1 in.
Width (Metric)48.5 cm
Width (Metric) Interior33 cm
Height (English)21 in.
Height (English) Interior6.8 in.
Height (Metric)53.3 cm
Height (Metric) Interior18 cm
Depth (English) Interior10 in.
Depth (Metric) Interior25 cm
Shipping Weight (English)134 lb.
Shipping Weight (Metric)60.8 kg
Electrical Requirements240 V 50/60 Hz
Voltage240 V
Amperage12.9 A
Frequency50/60 Hz
Unit SizeEach
F6018Full specifications
EachDigital single setpoint (A1)208 VCSA approved
-
Capacity (English)0.5 cu.ft.
Certifications/ComplianceCSA approved
DescriptionF6018
Capacity (Metric)14 L
Temperature (Metric)100°C to 1200°C
TypeSingle Setpoint
Power Consumption2325 w
ShelvesStainless Steel
Temperature ControlDigital single setpoint (A1)
Temperature Stability±0.3 at 1000°C
Temperature Uniformity±2.2 at 1000°C
Dimensions (D x W x H) Interior10 x 12.8 x 6.8 in. (25 x 33 x 18 cm)
Dimensions (L x W x H) Exterior20.1 x 19.1 x 21 in. (51 x 48.5 x 53.3 cm)
Length (English)20.1 in.
Length (Metric)51 cm
Width (English)19.1 in.
Width (Metric)48.5 cm
Width (Metric) Interior33 cm
Height (English)21 in.
Height (English) Interior6.8 in.
Height (Metric)53.3 cm
Height (Metric) Interior18 cm
Depth (English) Interior10 in.
Depth (Metric) Interior25 cm
Shipping Weight (English)134 lb.
Shipping Weight (Metric)60.8 kg
Electrical Requirements208 V 50/60 Hz
Voltage208 V
Amperage11.2 A
Frequency50/60 Hz
Unit SizeEach
F6020C-33Full specifications
EachDigital single setpoint with ramp and dwell (B1)240 VCSA approved, CE markedRequest A Quote--
Capacity (English)0.5 cu.ft.
Certifications/ComplianceCSA approved, CE marked
DescriptionF6020C-33
Capacity (Metric)14 L
Temperature (Metric)100°C to 1200°C
TypeSingle Setpoint
Power Consumption4400 w
ShelvesStainless Steel
Temperature ControlDigital single setpoint with ramp and dwell (B1)
Temperature Stability±1.5 at 1000°C
Temperature Uniformity±4.5 at 1000°C
Dimensions (D x W x H) Interior10 x 12.8 x 6.8 in. (25 x 33 x 18 cm)
Dimensions (L x W x H) Exterior20.1 x 19.1 x 21 in. (51 x 48.5 x 53.3 cm)
Length (English)20.1 in.
Length (Metric)51 cm
Width (English)19.1 in.
Width (Metric)48.5 cm
Width (Metric) Interior33 cm
Height (English)21 in.
Height (English) Interior6.8 in.
Height (Metric)53.3 cm
Height (Metric) Interior18 cm
Depth (English) Interior10 in.
Depth (Metric) Interior25 cm
Shipping Weight (English)134 lb.
Shipping Weight (Metric)60.8 kg
Electrical Requirements240 V 50/60 Hz
Voltage240 V
Amperage18.3 A
Frequency50/60 Hz
Unit SizeEach
F6020C-33-80Full specifications
EachDigital programmable with 4 programs (C1)240 VCSA approved, CE markedRequest A Quote--
Capacity (English)0.5 cu.ft.
Certifications/ComplianceCSA approved, CE marked
DescriptionF6020C-33-80
Capacity (Metric)14 L
Temperature (Metric)100°C to 1200°C
TypeMultiprogram
Power Consumption4400 w
ShelvesStainless Steel
Temperature ControlDigital programmable with 4 programs (C1)
Temperature Stability±1.5 at 1000°C
Temperature Uniformity±4.5 at 1000°C
Dimensions (D x W x H) Interior10 x 12.8 x 6.8 in. (25 x 33 x 18 cm)
Dimensions (L x W x H) Exterior20.1 x 19.1 x 21 in. (51 x 48.5 x 53.3 cm)
Length (English)20.1 in.
Length (Metric)51 cm
Width (English)19.1 in.
Width (Metric)48.5 cm
Width (Metric) Interior33 cm
Height (English)21 in.
Height (English) Interior6.8 in.
Height (Metric)53.3 cm
Height (Metric) Interior18 cm
Depth (English) Interior10 in.
Depth (Metric) Interior25 cm
Shipping Weight (English)134 lb.
Shipping Weight (Metric)60.8 kg
Electrical Requirements240 V 50/60 Hz
Voltage240 V
Amperage18.3 A
Frequency50/60 Hz
Unit SizeEach
F6028CFull specifications
EachDigital single setpoint with ramp and dwell (B1)208 VCSA approved
-
Capacity (English)0.5 cu.ft.
Certifications/ComplianceCSA approved
DescriptionF6028C
Capacity (Metric)14 L
Temperature (Metric)100°C to 1200°C
TypeSingle
Power Consumption4000 w
ShelvesStainless Steel
Temperature ControlDigital single setpoint with ramp and dwell (B1)
Temperature Stability±1.5 at 1000°C
Temperature Uniformity±4.5 at 1000°C
Dimensions (D x W x H) Interior10 x 12.8 x 6.8 in. (25 x 33 x 18 cm)
Dimensions (L x W x H) Exterior20.1 x 19.1 x 21 in. (51 x 48.5 x 53.3 cm)
Length (English)20.1 in.
Length (Metric)51 cm
Width (English)19.1 in.
Width (Metric)48.5 cm
Width (Metric) Interior33 cm
Height (English)21 in.
Height (English) Interior6.8 in.
Height (Metric)53.3 cm
Height (Metric) Interior18 cm
Depth (English) Interior10 in.
Depth (Metric) Interior25 cm
Shipping Weight (English)134 lb.
Shipping Weight (Metric)60.8 kg
Electrical Requirements208 V 50/60 Hz
Voltage208 V
Amperage19.2 A
Frequency50/60 Hz
Unit SizeEach
F6028C-80Full specifications
EachDigital programmable with 4 programs (C1)208 VCSA approvedRequest A Quote--
Capacity (English)0.5 cu.ft.
Certifications/ComplianceCSA approved
DescriptionF6028C-80
Capacity (Metric)14 L
Temperature (Metric)100°C to 1200°C
TypeMultiprogram
Power Consumption4000 w
ShelvesStainless Steel
Temperature ControlDigital programmable with 4 programs (C1)
Temperature Stability±1.5 at 1000°C
Temperature Uniformity±4.5 at 1000°C
Dimensions (D x W x H) Interior10 x 12.8 x 6.8 in. (25 x 33 x 18 cm)
Dimensions (L x W x H) Exterior20.1 x 19.1 x 21 in. (51 x 48.5 x 53.3 cm)
Length (English)20.1 in.
Length (Metric)51 cm
Width (English)19.1 in.
Width (Metric)48.5 cm
Width (Metric) Interior33 cm
Height (English)21 in.
Height (English) Interior6.8 in.
Height (Metric)53.3 cm
Height (Metric) Interior18 cm
Depth (English) Interior10 in.
Depth (Metric) Interior25 cm
Shipping Weight (English)134 lb.
Shipping Weight (Metric)60.8 kg
Electrical Requirements208 V 50/60 Hz
Voltage208 V
Amperage19.2 A
Frequency50/60 Hz
Unit SizeEach
F6030CMFull specifications
EachDigital programmable with 4 programs, 16 segments240 VCSA approved
-
Capacity (English)0.5 cu.ft.
Certifications/ComplianceCSA approved
DescriptionF6030CM
Capacity (Metric)14 L
Temperature (Metric)100°C to 1200°C
TypeMultisegment, Multiprogram
Power Consumption4400 w
ShelvesStainless Steel
Temperature ControlDigital programmable with 4 programs, 16 segments
Temperature Stability±0.2 at 1000°C
Temperature Uniformity±2.2 at 1000°C
Dimensions (D x W x H) Interior10 x 12.8 x 6.8 in. (25 x 33 x 18 cm)
Dimensions (L x W x H) Exterior20.1 x 19.1 x 21 in. (51 x 48.5 x 53.3 cm)
Length (English)20.1 in.
Length (Metric)51 cm
Width (English)19.1 in.
Width (Metric)48.5 cm
Width (Metric) Interior33 cm
Height (English)21 in.
Height (English) Interior6.8 in.
Height (Metric)53.3 cm
Height (Metric) Interior18 cm
Depth (English) Interior10 in.
Depth (Metric) Interior25 cm
Shipping Weight (English)134 lb.
Shipping Weight (Metric)60.8 kg
Electrical Requirements240 V 50/60 Hz
Voltage240 V
Amperage18.3 A
Frequency50/60 Hz
Unit SizeEach
F6030CM-33Full specifications
EachDigital programmable with 4 programs, 16 segments240 VCSA approved, CE markedRequest A Quote--
Capacity (English)0.5 cu.ft.
Certifications/ComplianceCSA approved, CE marked
DescriptionF6030CM-33
Capacity (Metric)14 L
Temperature (Metric)100°C to 1200°C
TypeMultisegment, Multiprogram
Power Consumption4400 w
ShelvesStainless Steel
Temperature ControlDigital programmable with 4 programs, 16 segments
Temperature Stability±1.5 at 1000°C
Temperature Uniformity±4.5 at 1000°C
Dimensions (D x W x H) Interior10 x 12.8 x 6.8 in. (25 x 33 x 18 cm)
Dimensions (L x W x H) Exterior20.1 x 19.1 x 21 in. (51 x 48.5 x 53.3 cm)
Length (English)20.1 in.
Length (Metric)51 cm
Width (English)19.1 in.
Width (Metric)48.5 cm
Width (Metric) Interior33 cm
Height (English)21 in.
Height (English) Interior6.8 in.
Height (Metric)53.3 cm
Height (Metric) Interior18 cm
Depth (English) Interior10 in.
Depth (Metric) Interior25 cm
Shipping Weight (English)134 lb.
Shipping Weight (Metric)60.8 kg
Electrical Requirements240 V 50/60 Hz
Voltage240 V
Amperage18.3 A
Frequency50/60 Hz
Unit SizeEach
F6038CMFull specifications
EachDigital programmable with 4 programs, 16 segments208 VCSA approved
-
Capacity (English)0.5 cu.ft.
Certifications/ComplianceCSA approved
DescriptionF6038CM
Capacity (Metric)14 L
Temperature (Metric)100°C to 1200°C
TypeMultisegment, Multiprogram
Power Consumption4000 w
ShelvesStainless Steel
Temperature ControlDigital programmable with 4 programs, 16 segments
Temperature Stability±1.5 at 1000°C
Temperature Uniformity±4.5 at 1000°C
Dimensions (D x W x H) Interior10 x 12.8 x 6.8 in. (25 x 33 x 18 cm)
Dimensions (L x W x H) Exterior20.1 x 19.1 x 21 in. (51 x 48.5 x 53.3 cm)
Length (English)20.1 in.
Length (Metric)51 cm
Width (English)19.1 in.
Width (Metric)48.5 cm
Width (Metric) Interior33 cm
Height (English)21 in.
Height (English) Interior6.8 in.
Height (Metric)53.3 cm
Height (Metric) Interior18 cm
Depth (English) Interior10 in.
Depth (Metric) Interior25 cm
Shipping Weight (English)134 lb.
Shipping Weight (Metric)60.8 kg
Electrical Requirements208 V 50/60 Hz
Voltage208 V
Amperage19.3 A
Frequency50/60 Hz
Unit SizeEach
Showing 10 of 10
  • Four heating elements are located on the chamber top, bottom and sides for enhanced temperature uniformity
  • Built-in vent port removes undesirable contaminants and moisture to extend the life of the element and unit
  • Rear of the chamber incorporates a 0.95cm (0.38in.) dia. port for monitoring chamber temperatures with independent measuring devices
  • Optional stainless steel shelf doubles load capacity (maximum temperature of 900°C)
  • Door safety switch stops power to heating elements when door opens
  • Thermocouple break protection cuts power to the heating elements, preventing a thermocouple failure runaway condition
  • Furnaces with B1, C1, and D1 control also use a mechanical overtemperature protection relay
Choice of Temperature Controllers
  • Digital single setpoint control (A1): Single display shows actual temperature or setpoint
  • Digital single setpoint control with single ramp to setpoint and a dwell (B1): Single display shows actual temperature or setpoint
  • Digital programmable control with one stored program of eight segments (C1)
  • Digital programmable control with four stored programs, 16 segments per program (D1): Controller includes RS-232 communications
Includes:
Models F6010 and F6018 include a cord and plug set.
Requires:
All except Models F6010 and F6018 require a conduit connection.
Warranty: 1 year
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.