CO2 and LN2 Backup Systems for Ultra-Low Temperature Freezers
Catalog number: FCO456LEV
Have Questions?
Thermo Scientific™
CO2 and LN2 Backup Systems for Ultra-Low Temperature Freezers
Catalog number: FCO456LEV
Have Questions?
Ensure optimal security for critical samples by using Thermo Scientific™ CO2 and LN2 Backup Systems for Ultra-Low Temperature Freezers, for use with Thermo Scientific™ TSX, Forma™, HERAfreeze™, Revco™ and TS-Series freezers.
Catalog Number
FCO456LEV
Unit Size
Each
Description
CO2 Backup System: maintains temperature down to -67°C with CO2
Type
CO2 Backup System
For Use With (Equipment)
Forma FDE, Revco RDE, HERAfreeze HDE, and Thermo Scientific TDE Series (400 box capacity and larger)
Revco™ ExF, DxF, CxF Series, TSE and TSD ULT Series
Unit Size
Each
Showing 10 of 34Show more
Features:
LN2 backup system maintains temperature down to -80°C with liquid nitrogen
CO2 backup system maintains temperature down to -67°C with CO2
Ordering Information:
Available as field- or factory-installed. Models for 300 box capacity models are standalone. Field-installed units must be installed by qualified professionals.