Informational:Inquire about OEM or Commercial Supply version of this product here.
Thermo Scientific™

FastDigest Psp1406I

Catalog number: FD0944
Have Questions?
Thermo Scientific™

FastDigest Psp1406I

Catalog number: FD0944
Have Questions?
Catalog Number
FD0944
Unit Size
20 reactions
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
FD094420 reactions
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
5'  A  A ↓C  G  T  T   3' 
3'  T  T  G  C ↑A  A   5' 

Thermo Scientific FastDigest Psp1406I restriction enzyme recognizes AA^CGTT site and cuts best at 37°C in 5–15 minutes using universal FastDigest Buffer. Isoschizomers: AclI, UbaN9I.

Thermo Scientific FastDigest Psp1406I is one of an advanced line of fast restriction enzymes that are all 100% active in the universal FastDigest and FastDigest Green reaction buffers.

The universal buffer allows rapid single-, double-, or multiple DNA digestion within 5–15 minutes eliminating any need for buffer change or subsequent DNA clean-up steps. See Reaction Conditions for FastDigest Enzymes for a table of enzyme activity, heat inactivation and incubation times for this and other FastDigest restriction enzymes. DNA modifying enzymes, such as Klenow Fragment, T4 DNA Ligase, alkaline phosphatases and T4 DNA Polymerase all have 100% activity in FastDigest Buffer. Therefore, enzymes for downstream applications can be directly added to the FastDigest reaction mix.

For additional convenience FastDigest Green Buffer includes a density reagent and two tracking dyes for direct loading of digestion reaction products on gels.

Short incubation times and optimal composition of the universal FastDigest Buffer eliminate star activity effects.

Features

• 100% activity of all FastDigest enzymes in the universal buffer
• 100% buffer compatibility with downstream applications
• Complete digestion in 5–15 minutes
• Direct loading on gels
• No star activity
• 176 FastDigest enzymes available

Applications

• Molecular cloning
• Restriction site mapping
• Genotyping
• Southern Blot
• Restriction fragment length polymorphism (RFLP)
• SNP analysis

Note: The FastDigest Green Buffer offers the same high performance in DNA digestion and downstream applications as the colorless FastDigest Buffer. For applications that require product analysis by fluorescence excitation (e.g. concentration measurements in UV light) the colorless FastDigest Buffer is recommended.

For methylation sensitivity, refer to product specifications.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Methylation Sensitivity
CpG Methylation-Sensitive, Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive
Enzyme
Psp1406I
Compatible Buffer
10X FastDigest Buffer/FastDigest Green Buffer
Sensitive to Heat Inactivation
Yes
Optimal Reaction Temperature
37°C
Type IIS RE
No
Quantity
20 μL (20 Reactions)
Product Type
Restriction Enzyme
Research Category
Traditional Cloning
Product Line
FastDigest™
Overhang
5' protruding ends
Inactivation Temperature
65°C

Contents & Storage

• FastDigest Psp1406I (20 μL)
• 10X FastDigest Buffer (1.0 mL)
• 10X FastDigest Green Buffer (1.0 mL)

Store at –20°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text