Informational:Inquire about OEM or Commercial Supply version of this product here.
Thermo Scientific™

FastDigest SchI

Catalog number: FD1374
Have Questions?
Thermo Scientific™

FastDigest SchI

Catalog number: FD1374
Have Questions?
Catalog Number
FD1374
Unit Size
100 reactions
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
FD1374100 reactions
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
5'  G  A  G  T  C  N5 3'
3'  C  T  C  A  G  N5 5'

Thermo Scientific FastDigest SchI restriction enzyme recognizes GAGTC(5/5)^ site and cuts best at 37°C in 5–15 minutes using universal FastDigest Buffer. Isoschizomers: MlyI, PleI, PpsI, SchI.

Thermo Scientific FastDigest SchI is one of an advanced line of fast restriction enzymes that are all 100% active in the universal FastDigest and FastDigest Green reaction buffers.

The universal buffer allows rapid single-, double-, or multiple DNA digestion within 5–15 minutes eliminating any need for buffer change or subsequent DNA clean-up steps. See Reaction Conditions for FastDigest Enzymes for a table of enzyme activity, heat inactivation and incubation times for this and other FastDigest restriction enzymes. DNA modifying enzymes, such as Klenow Fragment, T4 DNA Ligase, alkaline phosphatases and T4 DNA Polymerase all have 100% activity in FastDigest Buffer. Therefore, enzymes for downstream applications can be directly added to the FastDigest reaction mix.

For additional convenience FastDigest Green Buffer includes a density reagent and two tracking dyes for direct loading of digestion reaction products on gels.

Short incubation times and optimal composition of the universal FastDigest Buffer eliminate star activity effects.

Features

• 100% activity of all FastDigest enzymes in the universal buffer
• 100% buffer compatibility with downstream applications
• Complete digestion in 5–15 minutes
• Direct loading on gels
• No star activity
• 176 FastDigest enzymes available

Applications

• Molecular cloning
• Restriction site mapping
• Genotyping
• Southern Blot
• Restriction fragment length polymorphism (RFLP)
• SNP analysis

Note: The FastDigest Green Buffer offers the same high performance in DNA digestion and downstream applications as the colorless FastDigest Buffer. For applications that require product analysis by fluorescence excitation (e.g. concentration measurements in UV light) the colorless FastDigest Buffer is recommended.

For methylation sensitivity, refer to product specifications.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Methylation Sensitivity
Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive, Not CpG Methylation-Sensitive
Enzyme
SchI
Compatible Buffer
10X FastDigest Buffer/FastDigest Green Buffer
Sensitive to Heat Inactivation
Yes
Optimal Reaction Temperature
37°C
Type IIS RE
Yes
Quantity
100 μL (100 Reactions)
Product Type
Restriction Enzyme
Research Category
Traditional Cloning
Product Line
FastDigest™
Overhang
Blunt
Inactivation Temperature
80°C

Contents & Storage

• FastDigest SchI (100 μL)
• 10X FastDigest Buffer (1.0 mL)
• 10X FastDigest Green Buffer (1.0 mL)

Store at –20°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text