Informational:Inquire about OEM or Commercial Supply version of this product here.
Thermo Scientific™

FastDigest BseJI

Catalog number: FD1714
Have Questions?
Thermo Scientific™

FastDigest BseJI

Catalog number: FD1714
Have Questions?
Catalog Number
FD1714
Unit Size
200 reactions
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
FD1714200 reactions
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
5'  G  A  T  N  N ↓N  N  A  T  C   3' 
3'  C  T  A  N  N ↑N  N  T  A  G   5' 

Thermo Scientific FastDigest BseJI restriction enzyme recognizes GATNN^NNATC site and cuts best at 65°C in 5–15 minutes using universal FastDigest Buffer. Isoschizomers: BsaBI, Bse8I.

Thermo Scientific FastDigest BseJI is one of an advanced line of fast restriction enzymes that are all 100% active in the universal FastDigest and FastDigest Green reaction buffers.

The universal buffer allows rapid single-, double-, or multiple DNA digestion within 5–15 minutes eliminating any need for buffer change or subsequent DNA clean-up steps. See Reaction Conditions for FastDigest Enzymes for a table of enzyme activity, heat inactivation and incubation times for this and other FastDigest restriction enzymes. DNA modifying enzymes, such as Klenow Fragment, T4 DNA Ligase, alkaline phosphatases and T4 DNA Polymerase all have 100% activity in FastDigest Buffer. Therefore, enzymes for downstream applications can be directly added to the FastDigest reaction mix.

For additional convenience FastDigest Green Buffer includes a density reagent and two tracking dyes for direct loading of digestion reaction products on gels.

Short incubation times and optimal composition of the universal FastDigest Buffer eliminate star activity effects.

Features

• 100% activity of all FastDigest enzymes in the universal buffer
• 100% buffer compatibility with downstream applications
• Complete digestion in 5–15 minutes
• Direct loading on gels
• No star activity
• 176 FastDigest enzymes available

Applications

• Molecular cloning
• Restriction site mapping
• Genotyping
• Southern Blot
• Restriction fragment length polymorphism (RFLP)
• SNP analysis

Note: The FastDigest Green Buffer offers the same high performance in DNA digestion and downstream applications as the colorless FastDigest Buffer. For applications that require product analysis by fluorescence excitation (e.g. concentration measurements in UV light) the colorless FastDigest Buffer is recommended.

For methylation sensitivity, refer to product specifications.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Methylation Sensitivity
CpG Methylation-Sensitive, Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive
Enzyme
BseJI
Compatible Buffer
10X FastDigest Buffer/FastDigest Green Buffer
Sensitive to Heat Inactivation
No
Optimal Reaction Temperature
65°C
Type IIS RE
No
Quantity
200 μL (200 Reactions)
Product Type
Restriction Enzyme
Research Category
Traditional Cloning
Product Line
FastDigest™
Overhang
Blunt

Contents & Storage

• FastDigest BseJI (200 μL)
• 10X FastDigest Buffer (1.0 mL)
• 10X FastDigest Green Buffer (1.0 mL)

Store at –20°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text