Analyze records of which staff opened the freezer door, what time is was opened and for what duration with the Thermo Scientific™ Access Key Options for Ultra-Low Temperature Freezers.
Catalog Number
FFAC34567
Unit Size
Each
For Use With (Equipment)
Thermo Scientific™ TSU Series
Description
Access Key Option; Card access control w/five key cards - Field Installed
TSX Series, Forma 89000, HERAfreeze HLE, Revco RLE and TLE Series
Description
Factory-Installed access key option; Card access w/five key cards. All orders must accompany a TSX, Revco RLE, Forma 89000, TLE or HERAfreeze HLE Series ultra-low freezer. No stand-alone orders.
Factory-Installed access key option; Card access w/five key cards. All orders must accompany a TSX, Revco RLE, Forma 89000, TLE or HERAfreeze HLE Series ultra-low freezer. No stand-alone orders.
Type
Freezer Access Key
For Use With (Equipment)
TSX Series, Forma 89000, HERAfreeze HLE, Revco RLE and TLE Series
Field-Installed access key option; Card access w/five key cards. All orders must accompany a TSX, Revco RLE, Forma 89000, TLE or HERAfreeze HLE Series ultra-low freezer. No stand-alone orders.
Price (USD)
1,420.00
Availability
-
Full specifications
Quantity
Description
Field-Installed access key option; Card access w/five key cards. All orders must accompany a TSX, Revco RLE, Forma 89000, TLE or HERAfreeze HLE Series ultra-low freezer. No stand-alone orders.
TSX Series, Forma 89000, HERAfreeze HLE, Revco RLE and TLE Series
Factory-Installed access key option; Card access w/five key cards. All orders must accompany a TSX, Revco RLE, Forma 89000, TLE or HERAfreeze HLE Series ultra-low freezer. No stand-alone orders.
Factory-Installed access key option; Card access w/five key cards. All orders must accompany a TSX, Revco RLE, Forma 89000, TLE or HERAfreeze HLE Series ultra-low freezer. No stand-alone orders.
Type
Freezer Access Key
For Use With (Equipment)
TSX Series, Forma 89000, HERAfreeze HLE, Revco RLE and TLE Series
Field-Installed access key option; Card access w/five key cards. All orders must accompany a TSX, Revco RLE, Forma 89000, TLE or HERAfreeze HLE Series ultra-low freezer. No stand-alone orders.
1,420.00
-
Description
Field-Installed access key option; Card access w/five key cards. All orders must accompany a TSX, Revco RLE, Forma 89000, TLE or HERAfreeze HLE Series ultra-low freezer. No stand-alone orders.
Access key option; Card access w/five key cards. All orders must accompany a TSX Universal ultra-low freezer. No stand-alone orders.
Type
Factory-Installed
For Use With (Equipment)
TSX Universal
Unit Size
Each
Showing 7 of 7
Analyze records of which staff opened the freezer door, what time is was opened and for what duration with the Thermo Scientific™ Access Key Options for Ultra-Low Temperature Freezers.