Thermo Scientific™
Helios™ G4 PFIB HXe DualBeam™ for Semiconductors
Catalog number: HELIOSG4HXE
The Thermo Scientific™ Helios™ G4 PFIB HXe DualBeam System provides unique capabilities to enable damage-free delayering of 10nm semiconductor devices and advanced failure analysis of 3D packages, in addition to a wide range of other large area FIB processing applications.