Thermo Scientific™
Helios 5 Hydra CX DualBeam
Catalog number: HYDRACX
The Thermo Scientific™ Helios 5 Hydra CX DualBeam is part of the fourth generation of the industry-leading Helios DualBeam family. It combines the new, innovative multiple ion species PFIB column with the monochromated Thermo Scientific Elstar SEM Column to provide the most advanced focused ion- and electron-beam performance.