Invitrogen™

Image-iT™ Green Hypoxia Reagent

Catalog number: I14833
Have Questions?
Invitrogen™

Image-iT™ Green Hypoxia Reagent

Catalog number: I14833
Have Questions?
Catalog Number
I14833
Unit Size
5 vials
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
1 vial
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
I148335 vials
-
I148341 vial
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Image-iT Green Hypoxia Reagent is a novel, fixable fluorogenic compound for measuring hypoxia in live cells. It is non-fluorescent when live cells are in an environment with normal oxygen concentrations and becomes fluorescent when oxygen levels are decreased. Image-iT Green Hypoxia Reagent sustains its fluorescence when cells/tissue return to normal oxygen levels, allowing the cells/tissue to be fixed with minimal loss of fluorescent signal.

Features of Image-iT Green Hypoxia Reagent include:
• Indicates hypoxia in live cells by fluorescing in low oxygen environments
• Fixable—sustains fluorescent signal when cells/tissue return to normal oxygen levels
• Easy to use—just add to cell culture media and image

Image-iT Green Hypoxia Reagent is a fluorogenic compound that is live cell permeable and becomes fluorescent in environments with low oxygen concentrations. These properties makes it a highly useful tool for detecting cells and tissue under hypoxic conditions. Image-iT Green Hypoxia Reagent is a very sensitive oxygen detector. Unlike pimonidazole adducts that respond only to very low oxygen levels, Image-iT Green Hypoxia Reagent begins to fluoresce when atmospheric oxygen levels are less than 5%. It responds quickly to such environments, and the fluorescence is sustained after the oxygen levels return to normal. These properties make Image-iT Green Hypoxia Reagent an ideal tool for detecting hypoxic conditions around tumors, 3D cultures, spheroids, neurons, etc.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Type
Reagent
Product Line
Image-iT™
Quantity
5 Vials
Reagent Type
Hypoxia Reagent
Shipping Condition
Approved for shipment on Wet or Dry Ice

Contents & Storage

Store at -5 to -30°C. Protect from light.
  • Stable for at least 6 months after receiving.
  • Figures

    Documents & Downloads

    Certificates

      Frequently asked questions (FAQs)

      Citations & References

      Search citations by name, author, journal title or abstract text