Image-iT™ Cell Painting Kit
Image-iT™ Cell Painting Kit
Invitrogen™

Image-iT™ Cell Painting Kit

The Image-iT Cell Painting Kit conveniently provides six different fluorescent labels to assess cellular morphology by high content screening forRead more
Promo ImagePromotion
Promo code:RIRP25
Save on Imaging Essentials to Create Your Next Masterpiece
14% off your purchase of $499 or more on Invitrogen cell stains and mounting products Learn More
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
I65500Promo Image10 384- or 96-well plates
I65000Promo Image2 384- or 96-well plates
Catalog number I65500
Price (USD)
1,361.65
Online Exclusive
1,404.00
Save 42.35 (3%)
Each
In stock
Add to cart
Quantity:
10 384- or 96-well plates
Recurring order eligible. Learn more »
Price (USD)
1,361.65
Online Exclusive
1,404.00
Save 42.35 (3%)
Each
Add to cart
Ask our AI about this Product

The Image-iT Cell Painting Kit conveniently provides six different fluorescent labels to assess cellular morphology by high content screening for drug discovery. Cell painting is useful for identifying cellular phenotypic changes, mechanisms of action, toxicity testing, and cellular profiling during high-content drug screening. This image-based assay known as cell painting uses fluorescent multiplexing to paint the specific organelle with its own label within the cell.

Using different fluorophores, the Image-iT Cell Painting Kit allows for cellular image-based profiling to characterize a cell’s response while visualizing and analyzing the spatial organization of structures to assess any morphological changes after exposure to drug candidates and other perturbations. In addition, cell painting allows researchers to investigate the organization of proteins, cell viability, and proliferation or visualize DNA damage. Cytological profiling studies can be performed using cell painting to evaluate the effects of compounds on cellular morphology to help identify drug candidates in high content screening assays.

When running high-content screening and imaging assays, high image quality is essential. Several factors can impair image quality, such as using thick, uneven plates, which can lead to focusing differences between wells and image quality issues across a plate. We recommend using our Optical-Quality Imaging Plates (Cat. No. A58941) for superior image quality and resolution when used with fluorescence or luminescence readers. When combined with the CellInsight High Content Screening (HCS) Platform, these cell painting kit reagents provide a phenotypical readout to depict cell health. The kit is based on the publication by Bray et al. (2016) that identified the morphological profiling of cells using multiplexed fluorescent dyes. Image-iT Cell Painting kits can be purchased with enough reagents for either two or ten 384-well plates.

This kit includes the following reagents:
• Hoechst 34580, trihydrochloride, trihydrate
• Concanavalin A, Alexa Fluor 488 conjugate
• Wheat Germ Agglutinin, Alexa Fluor 555 conjugate
• Alexa Fluor 568 Phalloidin
• SYTO 14 Green Fluorescent Nucleic Acid Stain
• MitoTracker Deep Red FM

For additional information see our Cell Painting application note

Specifications
FormatKit
Label TypeFluorescent Dye
Quantity10 384- or 96-well plates
Sub Cellular LocalizationPlasma Membrane, Nucleus, Mitochondria, Cytoplasm, Cytoskeleton, Golgi, Endoplasmic Reticulum, Nucleolus
ColorMulticolor
For Use With (Application)High-content Screening (HCS), Morphological Cell Profiling, Multiparameter Screening, Fluorescent Labeling, 3D Labeling
For Use With (Equipment)Fluorescence Microscope, High Content Analysis Instrument, CellInsight CX7 Pro, CellInsight CX7 LZR Pro
Product LineImage-iT
Product TypeCell Painting Kit
Unit SizeEach
Contents & Storage
Sufficient for ten 384- or 96-well plates
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Figures

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
3163197Certificate of AnalysisMay 04, 2025I65000
2971002Certificate of AnalysisSep 05, 2024I65000
2871983Certificate of AnalysisMar 15, 2024I65000
2739500Certificate of AnalysisJan 23, 2024I65000
2739499Certificate of AnalysisSep 22, 2023I65000
5 results displayed, search above for a specific certificate

Safety Data Sheets

Citations & References (2)

Citations & References
Abstract
Cell Painting: a decade of discovery and innovation in cellular imaging.
Authors:Seal S,Trapotsi MA,Spjuth O,Singh S,Carreras-Puigvert J,Greene N,Bender A,Carpenter AE
Journal:Nature methods
PubMed ID:39639168
Modern quantitative image analysis techniques have enabled high-throughput, high-content imaging experiments. Image-based profiling leverages the rich information in images to identify similarities or differences among biological samples, rather than measuring a few features, as in high-content screening. Here, we review a decade of advancements and applications of Cell Painting, a ... More
Cell Painting, a high-content image-based assay for morphological profiling using multiplexed fluorescent dyes.
Authors:Bray MA,Singh S,Han H,Davis CT,Borgeson B,Hartland C,Kost-Alimova M,Gustafsdottir SM,Gibson CC,Carpenter AE
Journal:Nature protocols
PubMed ID:27560178
In morphological profiling, quantitative data are extracted from microscopy images of cells to identify biologically relevant similarities and differences among samples based on these profiles. This protocol describes the design and execution of experiments using Cell Painting, a morphological profiling assay multiplexing six fluorescent dyes imaged in five channels, to ... More
2 total citations

Share catalog number, name or link

1x1 image pixel for data collection