Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Chemical Name or Material | Chondroitin sulfate |
Color | White |
Recommended Storage | Ambient temperatures |
EINECS Number | 232-696-9 |
TSCA | Yes |
MDL Number | MFCD00146419 |
Quantity | 25 g |
Merck Index | 14,2214 |
Synonym | chondroitin sulfate, chondroitin sulphate, chonsurid, chondroitin sulfate c, chondroitin polysulfate, chondroitin sulfates, chondroitin 4-sulfate, chondroitin 6-sulfate, chondroitin, hydrogen sulfate, chondroitin sulfuric acid |
Solubility Information | Soluble in water. |
InChI Key | KXKPYJOVDUMHGS-OSRGNVMNSA-N |
SMILES | CC(=O)NC1C(C(C(OC1O)OS(=O)(=O)O)O)OC2C(C(C(C(O2)C(=O)O)O)O)O |
IUPAC Name | (2S,3S,4S,5R,6R)-6-[(2R,3R,4R,5R,6R)-3-acetamido-2,5-dihydroxy-6-sulfooxyoxan-4-yl]oxy-3,4,5-trihydroxyoxane-2-carboxylic acid |
PubChem CID | 24766 |
Percent Purity | ≥90% |
Physical Form | Powder |
Unit Size | Each |
Chemical Name or Material | Chondroitin sulfate |
Color | White |
Recommended Storage | Ambient temperatures |
EINECS Number | 232-696-9 |
TSCA | Yes |
MDL Number | MFCD00146419 |
Quantity | 1 g |
Merck Index | 14,2214 |
Synonym | chondroitin sulfate, chondroitin sulphate, chonsurid, chondroitin sulfate c, chondroitin polysulfate, chondroitin sulfates, chondroitin 4-sulfate, chondroitin 6-sulfate, chondroitin, hydrogen sulfate, chondroitin sulfuric acid |
Solubility Information | Soluble in water. |
InChI Key | KXKPYJOVDUMHGS-OSRGNVMNSA-N |
SMILES | CC(=O)NC1C(C(C(OC1O)OS(=O)(=O)O)O)OC2C(C(C(C(O2)C(=O)O)O)O)O |
IUPAC Name | (2S,3S,4S,5R,6R)-6-[(2R,3R,4R,5R,6R)-3-acetamido-2,5-dihydroxy-6-sulfooxyoxan-4-yl]oxy-3,4,5-trihydroxyoxane-2-carboxylic acid |
PubChem CID | 24766 |
Percent Purity | ≥90% |
Physical Form | Powder |
Unit Size | Each |
Chemical Name or Material | Chondroitin sulfate |
Color | White |
Recommended Storage | Ambient temperatures |
EINECS Number | 232-696-9 |
TSCA | Yes |
MDL Number | MFCD00146419 |
Quantity | 5 g |
Merck Index | 14,2214 |
Synonym | chondroitin sulfate, chondroitin sulphate, chonsurid, chondroitin sulfate c, chondroitin polysulfate, chondroitin sulfates, chondroitin 4-sulfate, chondroitin 6-sulfate, chondroitin, hydrogen sulfate, chondroitin sulfuric acid |
Solubility Information | Soluble in water. |
InChI Key | KXKPYJOVDUMHGS-OSRGNVMNSA-N |
SMILES | CC(=O)NC1C(C(C(OC1O)OS(=O)(=O)O)O)OC2C(C(C(C(O2)C(=O)O)O)O)O |
IUPAC Name | (2S,3S,4S,5R,6R)-6-[(2R,3R,4R,5R,6R)-3-acetamido-2,5-dihydroxy-6-sulfooxyoxan-4-yl]oxy-3,4,5-trihydroxyoxane-2-carboxylic acid |
PubChem CID | 24766 |
Percent Purity | ≥90% |
Physical Form | Powder |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Quantity | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
J60341.14 also known as J60341-14 | Each | 25 g | 247.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
J60341.03 also known as J60341-03 | Each | 1 g | 30.50 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
J60341.06 also known as J60341-06 | Each | 5 g | 63.70 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|