Sodium acetate, 1M aq. soln., pH 4.5, RNAse free
Sodium acetate, 1M aq. soln., pH 4.5, RNAse free
Sodium acetate, 1M aq. soln., pH 4.5, RNAse free
Sodium acetate, 1M aq. soln., pH 4.5, RNAse free
Thermo Scientific Chemicals

Sodium acetate, 1M aq. soln., pH 4.5, RNAse free

CAS: 127-09-3 | C2H3NaO2 | 82.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
250 mL
Catalog number J63669.AE
also known as J63669-AE
Price (USD)
61.40
Each
Estimated availability date 06-Jul-2025
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (USD)
61.40
Each
Add to cart
Specifications
CAS127-09-3
ChEBICHEBI:32954
Chemical Name or MaterialSodium acetate
Concentration or Composition (by Analyte or Components)1 M aq. soln.
Formula Weight82.03
View more
Ask our AI about this Product
Sodium Acetate is used as a buffer in the buffering range of pH 3.6-5.6. And also used in the purification and precipitation of nucleic acids, in protein crystallization, Staining of gels in protein gel electrophoresis and HPLC. Mordant dyeing; DNA microarray studies of E. coli response. Investigation of protein unfolding during reverse phase chromatography. Used in conjunction with sodium carbonate to enhance the activation of freeze-dried subtilisin Carlsberg in organic solvents. As a substrate of acetokinase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium Acetate is used as a buffer in the buffering range of pH 3.6-5.6. And also used in the purification and precipitation of nucleic acids, in protein crystallization, Staining of gels in protein gel electrophoresis and HPLC. Mordant dyeing; DNA microarray studies of E. coli response. Investigation of protein unfolding during reverse phase chromatography. Used in conjunction with sodium carbonate to enhance the activation of freeze-dried subtilisin Carlsberg in organic solvents. As a substrate of acetokinase.

Solubility
Fully miscible.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
Q20L563Certificate of AnalysisMay 17, 2025J63669.AK, J63669.AE
P19L568Certificate of AnalysisMar 27, 2025J63669.AK, J63669.AE
Y26K569Certificate of AnalysisDec 02, 2024J63669.AK, J63669.AE
X20K563Certificate of AnalysisOct 25, 2024J63669.AK, J63669.AE
U25K525Certificate of AnalysisAug 29, 2024J63669.AK, J63669.AE
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection