Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The Thermo Scientific Maxima First Strand cDNA Synthesis Kit for RT-qPCR, available with or without dsDNase, is a convenient system optimized for cDNA synthesis in two-step quantitative RT-PCR (RT-qPCR) applications. It provides a dramatically simplified workflow that combines genomic DNA elimination and cDNA synthesis into one-tube procedure. These cDNA Synthesis kits use Maxima Reverse Transcriptase (RT), an advanced enzyme derived by in vitro evolution of M-MuLV RT. The enzyme features high thermostability, robustness and increased cDNA synthesis rate compared to wild type M-MuLV RT.
Format | Kit |
GC-Rich PCR Performance | High |
Reaction Speed | 30 min. |
Technique | Reverse Transcription |
Optimal Reaction Temperature | 50°C to 55°C |
Reverse Transcriptase | Maxima |
Ribonuclease H Activity | Yes |
Shipping Condition | Dry Ice |
For Use With (Application) | Real Time PCR (qPCR) |
Final Product Type | First-Strand cDNA |
No. of Reactions | 200 Reactions |
Reaction Format | Separate components |
Reagent Type | Reverse Transcription |
Size (Final Product) | Up to 20 kb |
Starting Material | RNA |
Includes | Kit with dsDNase |
Unit Size | Each |
Format | Kit |
GC-Rich PCR Performance | High |
Reaction Speed | 30 min. |
Technique | Reverse Transcription |
Optimal Reaction Temperature | 50°C to 55°C |
Reverse Transcriptase | Maxima |
Ribonuclease H Activity | Yes |
Shipping Condition | Dry Ice |
For Use With (Application) | Real Time PCR (qPCR) |
Final Product Type | First-Strand cDNA |
No. of Reactions | 50 Reactions |
Reaction Format | Separate components |
Reagent Type | Reverse Transcription |
Size (Final Product) | Up to 20 kb |
Starting Material | RNA |
Includes | Kit only |
Unit Size | Each |
Format | Kit |
GC-Rich PCR Performance | High |
Reaction Speed | 30 min. |
Technique | Reverse Transcription |
Optimal Reaction Temperature | 50°C to 55°C |
Reverse Transcriptase | Maxima |
Ribonuclease H Activity | Yes |
Shipping Condition | Dry Ice |
For Use With (Application) | Real Time PCR (qPCR) |
Final Product Type | First-Strand cDNA |
No. of Reactions | 200 Reactions |
Reaction Format | Separate components |
Reagent Type | Reverse Transcription |
Size (Final Product) | Up to 20 kb |
Starting Material | RNA |
Includes | Kit only |
Unit Size | Each |
Format | Kit |
GC-Rich PCR Performance | High |
Reaction Speed | 30 min. |
Technique | Reverse Transcription |
Optimal Reaction Temperature | 50°C to 55°C |
Reverse Transcriptase | Maxima |
Ribonuclease H Activity | Yes |
Shipping Condition | Dry Ice |
For Use With (Application) | Real Time PCR (qPCR) |
Final Product Type | First-Strand cDNA |
No. of Reactions | 50 Reactions |
Reaction Format | Separate components |
Reagent Type | Reverse Transcription |
Size (Final Product) | Up to 20 kb |
Starting Material | RNA |
Includes | Kit with dsDNase |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Includes | No. of Reactions | Price (USD) | Availability | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
K1672 | Each | Kit with dsDNase | 200 Reactions | 1,286.65 Online price 1,488.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
K1641 | Each | Kit only | 50 Reactions | 357.65 Online price 420.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
K1642 | Each | Kit only | 200 Reactions | 1,060.65 Online price 1,252.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
K1671 | Each | Kit with dsDNase | 50 Reactions | 396.65 Online price 456.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
The Maxima First Strand cDNA Synthesis Kit for RT-qPCR is capable of reproducible cDNA synthesis from a wide range of total RNA amounts (1 pg to 5 μg) at elevated temperatures (42–65°C). The synthesis reaction can be completed in 15–30 minutes. Components of the Maxima First Strand cDNA Synthesis Kit for RT-qPCR are pre-mixed to save time and to reduce the possibility of pipetting errors.
The Maxima First Strand cDNA Synthesis Kit for RT-qPCR with dsDNase also contains a novel double-strand specific DNase engineered to remove contaminating genomic DNA from RNA preps in two minutes without damage to quality or quantity of RNA. Highly specific activity towards double-stranded DNA ensures that single-stranded DNA (such as cDNA and primers) is not cleaved and dsDNase treated RNA can be directly added to reverse transcription.
Features of the Maxima First Strand cDNA Synthesis Kit for RT-qPCR include:
Maxima First Strand cDNA Synthesis Kit with dsDNase contains:
Kit with dsDNase also contains
Applications
Additional information about reaction components
The Maxima Enzyme Mix contains Maxima Reverse Transcriptase and RiboLock RNase Inhibitor. The recombinant RiboLock RNase Inhibitor effectively protects RNA templates from degradation by RNases A, B, and C at temperatures up to 55°C.
5X Reaction Mix contains the remaining reaction components: reaction buffer, dNTPs, oligo(dT)18 and random hexamer primers. Nuclease-free water is provided for reaction set-up and dilution of sample RNA.
The absence of endo- and exo-deoxyribonucleases, ribonucleases, and phosphatases has been confirmed by appropriate quality tests.
• Maxima Enzyme Mix (Maxima Reverse Transcriptase and RiboLock RNase Inhibitor)
• 5X Reaction Mix
• dsDNase
• 10X dsDNase Buffer
• Nuclease-free water
Store at –20°C.