Informational:Inquire about OEM or Commercial Supply version of this product here.
Informational:Informational:New optimized magnetic bead solution for DNA cleanup. Learn more here.
Informational:Informational:Need Ethanol or Isopropanol for your nucleic acid isolation? Visit our Molecular Biology Grade Solvents page.
Thermo Scientific™

MagJET NGS Cleanup and Size Selection Kit

Catalog number: K2823
Have Questions?
Thermo Scientific™

MagJET NGS Cleanup and Size Selection Kit

Catalog number: K2823
Have Questions?
Catalog Number
K2823
Unit Size
10 x 96 preps
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
4 x 96 preps
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
10 preps
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
1 x 96 preps
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
K282310 x 96 preps
-
K28224 x 96 preps
-
K282810 preps
-
K28211 x 96 preps
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

Thermo Scientific MagJET NGS Cleanup and Size Selection kit is designed for efficient and robust DNA fragment library cleanup and size-selection from a variety of enzymatic reaction mixtures including PCR, ligation, adapter addition and DNA end-repair reaction mixes. The kit utilizes high-capacity MagJET paramagnetic bead technology that ensures efficient recovery of DNA fragment library within a desired fragment length range. The purified DNA fragments are free of any next-generation sequencing workflow inhibiting components, such as sequencing adapters, primer dimers, unincorporated nucleotides, enzymes or salts, and are ready-to-use in downstream NGS applications. No prior DNA fragment library purification is required before the cleanup and size-selection procedures.

The kit is suitable for cleanup or size selection of 5 ng to 5 μg of DNA fragment library. MagJET cleanup and adapter removal protocols can easily be automated for higher throughput on automatic platforms such as Thermo Scientific KingFisher Flex and Thermo Scientific KingFisher Duo.

Highlights
Robust—reliable cleanup and size selection from different reaction mixtures and buffers
Flexible—user-defined size-selection of exact DNA fragment range
NGS-grade purity—complete removal of NGS adapters, primers, nucleotides, enzymes and other reaction components inhibiting NGS workflow
High throughput compatible—easy to automate on the Thermo Scientific KingFisher magnetic particle processors for high-throughput sample handling

Applications
DNA fragment library cleanup and size-selection for downstream use in next generation sequencing workflows.

Includes
• MagJET Magnetic Beads
• Binding Solution
• Wash Buffer (concentrated)
• Elution Buffer
• DNA Fragment Mix

Related Products
MagJET NGS Cleanup and Size Selection Kit

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Format
Kit
Isolation Technology
Magnetic Bead
Sequencing Type
Genome and DNA Sequencing
Final Product Type
DNA
For Use With (Equipment)
Magnetic Stand, KingFisher™ mL, KingFisher™ Flex
Product Line
MagJET
Product Type
NGS Cleanup and Size Selection Kit
Quantity
10 x 96 Preps
Unit Size
10 x 96 preps

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text