Informational:Informational:New optimized magnetic bead solution for DNA cleanup. Learn more here.
Informational:Informational:Need Ethanol or Isopropanol for your nucleic acid isolation? Visit our Molecular Biology Grade Solvents page.
Thermo Scientific™

MagJET NGS Cleanup and Size Selection Kit

Catalog number: K2828
Have Questions?
Thermo Scientific™

MagJET NGS Cleanup and Size Selection Kit

Catalog number: K2828
Have Questions?
Catalog Number
K2828
Unit Size
10 preps
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
4 x 96 preps
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
1 x 96 preps
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
10 x 96 preps
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
K282810 preps
-
K28224 x 96 preps
-
K28211 x 96 preps
-
K282310 x 96 preps
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

Thermo Scientific MagJET NGS Cleanup and Size Selection kit is designed for efficient and robust DNA fragment library cleanup and size-selection from a variety of enzymatic reaction mixtures including PCR, ligation, adapter addition and DNA end-repair reaction mixes. The kit utilizes high-capacity MagJET paramagnetic bead technology that ensures efficient recovery of DNA fragment library within a desired fragment length range. The purified DNA fragments are free of any next-generation sequencing workflow inhibiting components, such as sequencing adapters, primer dimers, unincorporated nucleotides, enzymes or salts, and are ready-to-use in downstream NGS applications. No prior DNA fragment library purification is required before the cleanup and size-selection procedures.

The kit is suitable for cleanup or size selection of 5 ng to 5 μg of DNA fragment library. MagJET cleanup and adapter removal protocols can easily be automated for higher throughput on automatic platforms such as Thermo Scientific KingFisher Flex and Thermo Scientific KingFisher Duo.

Highlights
Robust—reliable cleanup and size selection from different reaction mixtures and buffers
Flexible—user-defined size-selection of exact DNA fragment range
NGS-grade purity—complete removal of NGS adapters, primers, nucleotides, enzymes and other reaction components inhibiting NGS workflow
High throughput compatible—easy to automate on the Thermo Scientific KingFisher magnetic particle processors for high-throughput sample handling

Applications
DNA fragment library cleanup and size-selection for downstream use in next generation sequencing workflows.

Includes
• MagJET Magnetic Beads
• Binding Solution
• Wash Buffer (concentrated)
• Elution Buffer
• DNA Fragment Mix

Related Products
MagJET NGS Cleanup and Size Selection Kit

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Isolation Technology
Magnetic Bead
Sample Type
Enzymatic Reactions (PCR, ligation, adapter addition, DNA end-repair reaction)
Elution Volume
10 to 50 μL
Final Product Type
DNA
For Use With (Equipment)
KingFisher™ Flex, KingFisher™ Duo Prime
For Use With (Application)
PCR, Southern blotting, sequencing, nucleic acid labeling, hybridization
High-throughput Compatibility
High-throughput Compatible
Quantity
10 Preps
Shipping Condition
Room Temperature
Yield
Up to 700 bp
Starting Material Amount
100 μL (5 ng to 5 μg DNA fragments)

Contents & Storage

• 1 mL DNA Fragment Mix; -20°C
• 110 μL MagJET Magnetic Beads; 4°C
• 8 mL Binding Buffer (concentrated); room temperature
• 3 mL Wash Buffer (concentrated); room temperature
• 2 x 1.5 mL Elution Buffer; room temperature

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text