Invitrogen™

MagicMark™ XP Western Protein Standard

Catalog number: LC5602X4
Have Questions?
Invitrogen™

MagicMark™ XP Western Protein Standard

Catalog number: LC5602X4
Have Questions?
Catalog Number
LC5602X4
Unit Size
4 x 250 μL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
250 µL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
50 µL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
LC5602X44 x 250 μL
-
LC5602250 µL
-
LC560350 µL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
MagicMark XP Western Protein Standard consists of nine recombinant proteins (20–220 kDa), each of which contains an IgG binding site. The IgG binding site binds the primary or secondary antibody used for detection of the target protein, allowing direct visualization of the standard on the western blot. The MagicMark XP Standard is compatible with most western kits and substrates (chemiluminescent, chromogenic, and fluorescent). The protein standard is supplied in a ready-to-use format for direct loading onto gels; no need to heat, reduce, or add sample buffer prior to use.

Compare and view all other protein standards and ladders ›

Applications
• Western blotting: detection of the nine unstained bands via the detection method used for the target protein. Compatible with chemiluminescent substrates and fluorescent secondary antibodies (not recommended for antibodies labeled with fluors in the 500–550 nm channel).
• Protein size estimation: protein sizing on SDS-polyacrylamide gels and on western blot using western blot detection method

Specifications

Detection Method
User defined detection system
Number of Markers
9
Product Type
Protein Standard
Ready to Load
Yes
Size Range
20 to 220 kDa
Stain Type
Unstained
Gel Compatibility
Bolt™ Bis-Tris Plus Gels, Novex™ Tricine Gels, Novex™ Tris-Glycine Gels, NuPAGE™ Bis-Tris Gels, NuPAGE™ Tris-Acetate Gels, SDS-PAGE Gels
Kit Contents
4 x 250 μL of ready-to-use standard mixture is provided in a plastic vial; Loading buffer consists of 125mM Tris-HCl (pH 6.8), 10mM DTT, 17.4% Glycerol, 3% SDS, and 0.025% Bromophenol Blue
Molecular Weight (g/mol)
220, 120, 100, 80, 60, 50, 40, 30, 20 kDa
Product Line
MagicMark™
Quantity
4 x 250 μL
Shipping Condition
Dry Ice
System Type
Western Blotting, SDS-PAGE

Contents & Storage

4 x 250 μL of ready-to-use standard mixture provided in plastic vials. Loading buffer consists of 125 mM Tris-HCl (pH 6.8), 10 mM DTT, 17.4% Glycerol, 3% SDS, and 0.025% Bromophenol Blue. Store at -20°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text