Applied Biosystems™

CapSure™ Macro LCM Caps

Catalog number: LCM0211
Have Questions?
Applied Biosystems™

CapSure™ Macro LCM Caps

Catalog number: LCM0211
Have Questions?
Catalog Number
LCM0211
Unit Size
48 caps
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
240 caps
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
LCM021148 caps
-
LCM0212240 caps
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

Specially designed for use with the Arcturus™ Laser Capture Microdissection (LCM) instruments, CapSure™ HS and CapSure Macro LCM Caps enable the precise and rapid extraction of pure populations of cells from heterogeneous sample preparations. CapSure Caps may be used to isolate individual cells or thousands of cells on a single cap.

Key product features:
• Gentle application – non-damaging microdissection preserves the integrity of the captured material
• Versatile extraction – capture both large and small amounts of cells

Ensure the Integrity of Microdissected Cells for Downstream Molecular Analysis
All CapSure™ LCM Caps have a patented transfer film bonded to the lower cap surface. Using the Arcturus Microdissection Systems an infrared laser pulses through the top of the cap during LCM and interacts with the transfer film, which then melts and bonds to the cells or regions of interest. The film absorbs the laser radiation – instead of the tissue or cell sample – creating a gentle, non-damaging microdissection that preserves the integrity of the captured material.

Extract Large Amounts of Cells with CapSure™ Macro LCM Caps
CapSure™ Macro LCM Caps provide the maximum area for capturing several thousand cells on a single cap. This is particularly useful for downstream applications that require large numbers of cells to obtain enough material for analysis, such as some protein analysis techniques. The CapSure™ Macro LCM Caps fit directly onto 0.5 ml microcentrifuge tubes for extraction of biological molecules from captured cells.

Extract Small Numbers of Cells with High Sensitivity CapSure™ HS Caps
CapSure™ HS LCM Caps enable the highly sensitive extraction and detection of biological molecules from small numbers of cells. CapSure™ HS LCM Caps contain ridges at the bottom that keep the active capture area of the cap twelve microns off the surface of the sample. This separation ensures the precise capture of up to several hundreds of cells. In addition, the CapSure™ LCM Caps work with ExtracSure devices that minimize the dilution of biological molecules extracted from captured cells. This optimizes the recovery of these molecules for downstream molecular analysis.

For Research Use Only. Not for use in diagnostics procedures.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Type
Caps
Product Line
CapSure
For Use With (Equipment)
ArcturusXT™ LCM Instrument
Includes
Capsure Macro LCM Caps
Material
LCM
Quantity
48 Caps
Shipping Condition
Room Temperature

Contents & Storage

CapSure Macro LCM Caps. Store at room temperature

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text