Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Reconstitute at 0.5 mg/mL in sterile PBS.
This gene encodes a transmembrane protein with two extracellular Kunitz domains that inhibits a variety of serine proteases. The protein inhibits HGF activator which prevents the formation of active hepatocyte growth factor. This gene is a putative tumor suppressor, and mutations in this gene result in congenital sodium diarrhea. Multiple transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ45571; HAI-2; Hepatocyte growth factor activator inhibitor type 2; Kunitz-type protease inhibitor 2; Placental bikunin; serine peptidase inhibitor, Kunitz type, 2; serine protease inhibitor, Kunitz type, 2; testicular tissue protein Li 183
Gene Aliases: DIAR3; HAI-2; HAI2; KOP; PB; SPINT2
UniProt ID: (Human) O43291
Entrez Gene ID: (Human) 10653
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support