Invitrogen™

MagniSort™ Streptavidin Negative Selection Beads

Catalog number: MSNB-6002-74
Have Questions?
Invitrogen™

MagniSort™ Streptavidin Negative Selection Beads

Catalog number: MSNB-6002-74
Have Questions?
Catalog Number
MSNB-6002-74
Unit Size
200 tests
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
25 tests
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
MSNB-6002-74200 tests
-
MSNB-6002-7125 tests
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
MagniSort™ Streptavidin Negative Selection Beads are designed for the magnetic separation of cells by negative selection. Undesired cells are bound by a cocktail of biotinylated antibodies and then streptavidin coated magnetic beads. When placed in a magnetic field, the undesired cells are held in place and the desired cells can be separated by decanting. The clone and concentration of biotinylated antibody to be used to label the undesired cells, as well as the concentration of MagniSort™ Streptavidin Negative Selection Beads must be determined empirically. Please refer to the protocol for details on how to optimize both the antibody or antibody cocktail and MagniSort™ Streptavidin Negative Selection Beads.

For depletion of cells using a single antibody, we recoomend the MagniSort™ Streptavidin Positive Selection Beads (cat. MSPB-6003) and protocol.

Reported Application
Magnetic Cell Separation

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Quantity
200 Tests
Form
Liquid
Ligand
Streptavidin
Product Type
Streptavidin Conjugate (Fluorescent)
Target Molecule
Biotin
Unit Size
200 tests

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text