Invitrogen™

MagniSort™ Streptavidin Positive Selection Beads

Catalog number: MSPB-6003-71
Have Questions?
Invitrogen™

MagniSort™ Streptavidin Positive Selection Beads

Catalog number: MSPB-6003-71
Have Questions?
Catalog Number
MSPB-6003-71
Unit Size
25 tests
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
200 tests
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
MSPB-6003-7125 tests
-
MSPB-6003-74200 tests
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
MagniSort™ Streptavidin Positive Selection Beads are designed for the magnetic separation of cells by positive selection. Desired cells are bound by biotinylated antibody and then streptavidin-coated magnetic beads. When placed in a magnetic field, the desired cells are held in place and the undesired cells can be separated by decanting. The clone and concentration of biotinylated antibody to be used to label the desired cells, as well as the concentration of MagniSort™ Streptavidin Positive Selection Beads must be determined empirically. Please refer to the protocol for details on how to optimize both the antibody and MagniSort™ Streptavidin Positive Selection Beads.

The MagniSort™ Streptavidin Positive Selection Beads may also be used to deplete undesired cells using a single antibody. For depletion of cells using multiple antibodies, we recommend the MagniSort™ Streptavidin Negative Selection Beads (cat. MSNB-6002) and protocol.

Reported Application
Magnetic Cell Separation

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Quantity
25 Tests
Form
Liquid
Ligand
Streptavidin
Product Type
Streptavidin Conjugate (Fluorescent)
Target Molecule
Biotin

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text