AmpliTaq™ DNA Polymerase with Buffer I or Buffer II
AmpliTaq™ DNA Polymerase with Buffer I or Buffer II
Applied Biosystems™

AmpliTaq™ DNA Polymerase with Buffer I or Buffer II

AmpliTaq DNA Polymerase is a 94 kDa, thermostable, recombinant DNA polymerase obtained by expression of a modified form of the Thermus aquaticus (Taq) DNA polymerase gene in E. coli.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberIncludesQuantity
N8080160Buffer I250 Units
N8080161Buffer II250 Units
N8080172Buffer II1000 Units
N8080153Buffer II3,000 units
N8080156Buffer II5,000 units
N8080186Buffer II25,000 units
N8080152Buffer I3,000 units
N8080155Buffer I5,000 units
N8080171Buffer I1,000 units
N8080185Buffer I25,000 Units
Catalog number N8080160
Price (EUR)
259,00
Each
-
Add to cart
Includes:
Buffer I
Quantity:
250 Units
Request bulk or custom format
Price (EUR)
259,00
Each
Add to cart
Ask our AI about this Product
AmpliTaq DNA Polymerase is a 94-kDa, thermostable, recombinant DNA polymerase obtained by expression of a modified form of the Thermus aquaticus (Taq) DNA polymerase gene in E. coli. It is the most thoroughly characterized enzyme available for the PCR process, and its recombinant nature and purification method provide unparalleled purity and reproducibility, vial-to-vial and lot-to-lot.

AmpliTaq DNA Polymerase is available with GeneAmp10X PCR Buffer I or with GeneAmp 10X PCR Buffer II and MgCl2 Solution.

Reliable and robust PCR

The thermal activity profile of AmpliTaq DNA Polymerase is good for PCR applications because its optimal activity is in the same range at which stringent annealing of primers occurs (55–75°C). The enzyme's half-life is ∼40 minutes at 95°C, providing thermostability that meets the requirements of most difficult PCR applications.

Features

  • The most thoroughly characterized enzyme available for the PCR process
  • Excellent thermal activity profile makes it good for PCR applications
  • QC-tested to guarantee reproducible results

Notes

  • For superior PCR performance, we recommend DreamTaq DNA Polymerase.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration5 U/μL
Exonuclease Activity5' - 3'
Fidelity (vs. Taq)1X
FormatStand-alone enzyme
Hot StartNo
IncludesBuffer I
No. of Reactions200 Reactions
Overhang3'-A
PolymeraseAmpliTaq DNA Polymerase
Product TypeDNA Polymerase
Quantity250 Units
Reaction FormatSeparate Components
Shipping ConditionDry Ice
Size (Final Product)5 kb or less
Starting MaterialDNA
For Use With (Application)Standard PCR
Reaction SpeedStandard
Unit SizeEach
Contents & Storage
• AmpliTaq DNA Polymerase (5 U/μL), 50 μL
• GeneAmp 10X PCR Buffer, 1.5 mL

Store at -20°C.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Safety Data Sheets

Other products to consider



Share catalog number, name or link

1x1 image pixel for data collection