MAS™ Omni·CORE Controls
MAS™ Omni·CORE Controls
Thermo Scientific™

MAS™ Omni·CORE Controls

Consolidate general chemistry and serum protein QC processing into a single product offering three distinct levels.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLevel
OCR-1011
OCR-2022
OCR-3033
Catalog number OCR-101
Price (USD)
391.00
Each
In stock
Add to cart
Level:
1
Recurring order eligible. Learn more »
Price (USD)
391.00
Each
Add to cart
Ask our AI about this Product
Consolidate general chemistry and serum protein QC processing into a single product offering three distinct levels with Thermo Scientific™ MAS™ Omni·CORE Controls. Value assignment is provided for key integrated instrument systems.
  • Three distinct levels
  • Liquid, ready-to-use
  • Human-serum based matrix
  • Assayed values available for most analyzers
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
DescriptionOmni CORE Control, Assayed
Detectable AnalytesAcetaminophen, Albumin, Alkaline Phosphatase, alpha-1-Acid Glycoprotein, alpha-1-Antitrypsin, alpha-2-Macroglobulin, ALT, Amikacin, Amylase, Amylase (Pancreatic), Antistreptolysin O (ASO), Apolipoprotein A1, Apolipoprotein B, AST, beta-2-microglobulin (B2M), Bile Acids, Bilirubin Conjugated, Bilirubin Direct (DBIL), Bilirubin Neonatal, Bilirubin Total (TBIL), Bilirubin Unconjugated, BUN, C3 Complement, C4 Complement, Caffeine, Calcium, Carbamazepine, Ceruloplasmin, Chloride, Cholesterol, Creatine Kinase (CK), CO2, Copper, Cortisol, C-Reactive Protein (CRP), Creatinine, Digoxin, Disopyramide, Ethanol, Ethosuximide, Ferritin, Gentamicin, GGT, Glucose, Glutamate Dehydrogenase (GLDH), Haptoglobin, HDL, IgA, IgE, IgG, IgM, Iron, Iron Binding Capacity (IBC), Iron Binding Capacity, Total (TIBC), Iron Binding Capacity, Unsaturated (UIBC), Lactic Acid, LDH, LDL, Lidocaine, Lipase, Lipoprotein (a), Lithium, Magnesium, Methotrexate, N-acetylprocainamide (NAPA), Osmolality, Phenobarbital, Phenytoin, Phospholipids, Phosphorus, Potassium, Prealbumin, Primidone, Procainamide, Protein Electrophoresis, Pseudocholinesterase (PCHE), Quinidine, Rheumatoid Factor, Salicylate (SALY), Sodium, Thyroxine (T4 Total), Theophylline, Tobramycin, Total Protein, Transferrin (TRF), Tricyclic Antidepressants, Triglycerides, Thyroid Stimulating Hormone (TSH), T-uptake, Uric Acid, Valproic Acid, Vancomycin, Zinc
Sample TypeSerum
TypeControl
Level1
Packaging6 Pk.
Quantity5 mL
Shelf Life36 Months at -25° to -15°C
Stability2° to 8°C
Unit SizeEach
Contents & Storage
36 month shelf life from date of manufacture when stored at -25° to -15°C; 30-day open-vial stability when stored at 2° to 8°C
lab-link

LabLink xL quality assurance software

Real-time, web-based data processing, peer data comparison, reports and documentation.

View MAS QC package inserts

Log in to LabLink xL

Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
OCR27071_073127Certificate of AnalysisMar 20, 2025OCR-101
OCR27072_073127Certificate of AnalysisMar 20, 2025OCR-202
OCR27073_073127Certificate of AnalysisMar 20, 2025OCR-303
OCR27043_043027Certificate of AnalysisFeb 21, 2025OCR-303LSG, OCR-303
OCR27083_083127Certificate of AnalysisFeb 21, 2025OCR-303LSG, OCR-303
5 results displayed, search above for a specific certificate

Share catalog number, name or link

1x1 image pixel for data collection