ProSightPD Software
Catalog number: OPTON-30918
Have Questions?
ProSightPD Software
Catalog number: OPTON-30918
Have Questions?
Perform comprehensive analysis of top-down proteomics data with Thermo Scientific™ ProSightPD™ software. ProSightPD features robust proteomic search engines for proteoform characterization, including proteoforms with PTMs or truncations. ProSightPD software accommodates proteomic data generated by fragmentation techniques present in any Thermo Scientific mass spectrometer. ProSightPD software is the only proteomics software that allows the user to easily search MS/MS data against proteome databases containing the complete biological complexity found in UniProt.
Catalog Number
OPTON-30918
Unit Size
Each
Description
ProSightPD software.
Price (USD)
Full specifications
TypeSoftware
Software2
DescriptionProSightPD software.
For Use With (Equipment)Mass Spectrometers
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Thermo Scientific ProSightPD software.
Price (USD)
Full specifications
TypeSoftware
Software2
DescriptionThermo Scientific ProSightPD software.
For Use With (Equipment)Mass Spectrometers
Unit SizeEach
Catalog Number
Unit Size
Each
Description
ProSightPD 4.2 software
Price (USD)
Full specifications
TypeSoftware
Software4.2
Description
ProSightPD 4.2 software
For Use With (Equipment)
Mass Spectrometers
IncludesThermo Scientific ProSight PD 4.2 software
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Upgrade ProSightPD 4.1 to ProSightPD 4.2
Price (USD)
Full specifications
TypeSoftware
Software
4.2
Description
Upgrade ProSightPD 4.1 to ProSightPD 4.2
For Use With (Equipment)
Mass Spectrometers
Includes
Thermo Scientific ProSight PD 4.2 software
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Upgrade ProSightPD before 4.1 to ProSightPD 4.2
Price (USD)
Full specifications
TypeSoftware
Software
4.2
Description
Upgrade ProSightPD before 4.1 to ProSightPD 4.2
For Use With (Equipment)
Mass Spectrometers
Includesp>Thermo Scientific ProSight PD 4.2 software
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Upgrade ProSightPD 4.2 from ProSightPC
Price (USD)
Full specifications
TypeSoftware
Software
4.2
Description
Upgrade ProSightPD 4.2 from ProSightPC
For Use With (Equipment)
Mass Spectrometers
Includes
Thermo Scientific ProSight PD 4.2 software
Unit SizeEach
Showing 6 of 6
Catalog NumberSpecificationsUnit SizeDescriptionPrice (USD)
OPTON-30918Full specifications
EachProSightPD software.Request A Quote
TypeSoftware
Software2
DescriptionProSightPD software.
For Use With (Equipment)Mass Spectrometers
Unit SizeEach
OPTON-30903Full specifications
EachThermo Scientific ProSightPD software.Request A Quote
TypeSoftware
Software2
DescriptionThermo Scientific ProSightPD software.
For Use With (Equipment)Mass Spectrometers
Unit SizeEach
OPTON-31090Full specifications
Each
ProSightPD 4.2 software
Request A Quote
TypeSoftware
Software4.2
Description
ProSightPD 4.2 software
For Use With (Equipment)
Mass Spectrometers
IncludesThermo Scientific ProSight PD 4.2 software
Unit SizeEach
OPTON-31091Full specifications
Each
Upgrade ProSightPD 4.1 to ProSightPD 4.2
Request A Quote
TypeSoftware
Software
4.2
Description
Upgrade ProSightPD 4.1 to ProSightPD 4.2
For Use With (Equipment)
Mass Spectrometers
Includes
Thermo Scientific ProSight PD 4.2 software
Unit SizeEach
OPTON-31092Full specifications
Each
Upgrade ProSightPD before 4.1 to ProSightPD 4.2
Request A Quote
TypeSoftware
Software
4.2
Description
Upgrade ProSightPD before 4.1 to ProSightPD 4.2
For Use With (Equipment)
Mass Spectrometers
Includesp>Thermo Scientific ProSight PD 4.2 software
Unit SizeEach
OPTON-31093Full specifications
Each
Upgrade ProSightPD 4.2 from ProSightPC
Request A Quote
TypeSoftware
Software
4.2
Description
Upgrade ProSightPD 4.2 from ProSightPC
For Use With (Equipment)
Mass Spectrometers
Includes
Thermo Scientific ProSight PD 4.2 software
Unit SizeEach
Showing 6 of 6
Broad Functionality
A fast and flexible proteomics search engine for both targeted and high-throughput protein identification, designed specifically for top-down proteomics data.
  • Includes five different database search modes – Sequence Tag, Single Protein, and Gene Restricted search-for exceptional intact protein characterization.
  • Performs false discovery rate calculations at the protein, isoform, proteoform, and proteoform hit level.
  • Processes many different types of fragmentation data, including data generated using ETD, HCD, EThcD, UVPD, ECD, IRMPD, or CID.
  • Enables merging of graphical fragment maps, even those with different ion types.
  • Includes proprietary ProSight containing all known post-translational modifications (PTMs), alternative splicing events and single nucleotide polymorphisms (SNPs).
  • Offers more robust precursor mass analysis, with FAIMS and PTR data now readily analyzable using ProSightPD software. Both high- and low-resolution data can be processed with the Xtract and ReSpect algorithms embedded in ProSightPD software, allowing analysis of masses from a wide range of masses (∼1 kDa to >150 kDa).
  • Imports XML and FASTA databases and shotgun annotates them with all possible modifications.
  • Allows users to experience unparalleled, versatile visualization for reviewing search results and customizing modifications to find better result fits.