Thermo Scientific™

Triple Wrap Sterile Pack w/VHP Indicator Tryptone Soya Agar

Catalog number: PO5500B
Have Questions?
Thermo Scientific™

Triple Wrap Sterile Pack w/VHP Indicator Tryptone Soya Agar

Catalog number: PO5500B
Have Questions?
Experience next-level cleanroom confidence with Thermo Scientific™ Triple Wrap Sterile Pack Tryptone Soya Agar plates, complete with VHP Indicator.
 
Catalog Number
PO5500B
Unit Size
Each
Description
Triple Wrap Sterile Pack w/VHP Indicator Tryptone Soya Agar
dimFormat
Quantity
100/Pk.
Manufacturing Origin
Germany
Price (USD)
Full specifications
ColorIvory, transparent
DescriptionTriple Wrap Sterile Pack w/VHP Indicator Tryptone Soya Agar
Format90mm Monoplate
Manufacturing OriginGermany
Product TypeAgar
Fill Weight25.0g ± 5%
Packaging TypeTriple Wrapped - Irradiated
Quantity100/Pk.
Shelf Life43 Weeks
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Triple Wrap Sterile Pack w/VHP Indicator Tryptone Soya Agar
dimFormat
Quantity
100/Pk.
Manufacturing Origin
Germany
Price (USD)
Full specifications
ColorIvory, transparent
DescriptionTriple Wrap Sterile Pack w/VHP Indicator Tryptone Soya Agar
Format55mm Contact Plate
Manufacturing OriginGermany
Product TypeAgar
Fill Weight13.0g ± 0.5g
Packaging TypeTriple Wrapped - Irradiated
Quantity100/Pk.
Shelf Life43 Weeks
Unit SizeEach
Showing 2 of 2
Catalog NumberSpecificationsUnit SizeDescriptiondimFormatQuantityManufacturing OriginPrice (USD)
PO5500BFull specifications
EachTriple Wrap Sterile Pack w/VHP Indicator Tryptone Soya Agar100/Pk.GermanyRequest A Quote
ColorIvory, transparent
DescriptionTriple Wrap Sterile Pack w/VHP Indicator Tryptone Soya Agar
Format90mm Monoplate
Manufacturing OriginGermany
Product TypeAgar
Fill Weight25.0g ± 5%
Packaging TypeTriple Wrapped - Irradiated
Quantity100/Pk.
Shelf Life43 Weeks
Unit SizeEach
PO5510DFull specifications
EachTriple Wrap Sterile Pack w/VHP Indicator Tryptone Soya Agar100/Pk.GermanyRequest A Quote
ColorIvory, transparent
DescriptionTriple Wrap Sterile Pack w/VHP Indicator Tryptone Soya Agar
Format55mm Contact Plate
Manufacturing OriginGermany
Product TypeAgar
Fill Weight13.0g ± 0.5g
Packaging TypeTriple Wrapped - Irradiated
Quantity100/Pk.
Shelf Life43 Weeks
Unit SizeEach
Showing 2 of 2
Achieve confidence in your results and ensure optimum media fertility using plates with a Vapour-phase Hydrogen Peroxide (VHP) exposure indicator, designed to confirm the integrity of the wrapping material and seal during VHP exposure.

Three protective layers reduce risk of contamination:
  • Primary film with moisture control patch
  • Secondary film VHP barrier with immobilized desiccant sachet
  • Tertiary film dust cover
The incorporation of novel elements enhance moisture control by permitting the passage and absorption of excess moisture

Using plates with an improved shelf life and room temperature storage offers a cost-effective solution, minimizing new batch validation and refrigeration costs

Not all products are available for sale in all territories. Please inquire.

Contents & Storage

2 to 25°C

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text