Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Color | Ivory, transparent |
Description | Triple Wrap Sterile Pack w/VHP Indicator Tryptone Soya Agar |
Format | 55mm Contact Plate |
Manufacturing Origin | Germany |
Product Type | Agar |
Fill Weight | 13.0g ± 0.5g |
Packaging Type | Triple Wrapped - Irradiated |
Quantity | 100/Pk. |
Shelf Life | 43 Weeks |
Unit Size | Each |
Color | Ivory, transparent |
Description | Triple Wrap Sterile Pack w/VHP Indicator Tryptone Soya Agar |
Format | 90mm Monoplate |
Manufacturing Origin | Germany |
Product Type | Agar |
Fill Weight | 25.0g ± 5% |
Packaging Type | Triple Wrapped - Irradiated |
Quantity | 100/Pk. |
Shelf Life | 43 Weeks |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | dimFormat | Quantity | Manufacturing Origin | Price (USD) | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
PO5510D | Each | Triple Wrap Sterile Pack w/VHP Indicator Tryptone Soya Agar | 100/Pk. | Germany | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
PO5500B | Each | Triple Wrap Sterile Pack w/VHP Indicator Tryptone Soya Agar | 100/Pk. | Germany | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|