See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
Home
Shop All Products
Process Controllers and Analyzers
Process Mass Spectrometers
ProLab Benchtop Mass Spectrometer
Thermo Scientific™
ProLab Benchtop Mass Spectrometer
Catalog number:
PROLBANLYZ
Related applications:
Mass Spectrometry
Have Questions?
Contact Us
Thermo Scientific™
ProLab Benchtop Mass Spectrometer
Catalog number:
PROLBANLYZ
Related applications:
Mass Spectrometry
Have Questions?
Contact Us
This product has been discontinued however we continue to provide services and support for this product.
Catalog Number
PROLBANLYZ
Unit Size
-
Product Type
Benchtop Mass Spectrometer
Price (USD)
Contact Us
Full specifications
Product Type
Benchtop Mass Spectrometer
Unit Size
-
Save to list
Showing 1 of 1
Catalog Number
Specifications
Unit Size
Product Type
Price (USD)
PROLBANLYZ
Full specifications
-
Benchtop Mass Spectrometer
Contact Us
Product Type
Benchtop Mass Spectrometer
Unit Size
-
Showing 1 of 1
Save to list
Features:
Single or triple filter quadrupole
Open or enclosed ion source
Simple capillary, leak valve, capillary or UHP all metal atmospheric inlet
Corrosive pump option (Advanced)
Temperature controlled bake out and inlet heating (Advanced)
10 ppb to 100% measurement range (Advanced)
Recommended for:
Catalysis research
Thermal analysis
Semiconductor PFC monitoring
Gas purity measurements
Fermentation research
Pilot plant gas analysis
Transient kinetics studies
TPD
Evolved gas analysis
Air separation plant troubleshooting
Figures
Documents & Downloads
Certificates
Search by lot number or partial lot number
Search
Request a Certificate