Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Assay | RNA Quantitation, broad range |
Detection Method | Fluorescence |
Excitation/Emission | 644/673 |
For Use With (Equipment) | Microplate Reader |
No. of Reactions | 1000 Reactions (200 μL Assay Volume) |
Quantitation Range | 20 to 1000 ng |
Product Line | Quant-iT |
Shipping Condition | Room Temperature |
Unit Size | 1,000 assays |
Assay | RNA Quantitation |
Detection Method | Fluorescence |
For Use With (Equipment) | Microplate Reader |
No. of Reactions | 1000 Reactions (200 μL Assay Volume) |
Quantitation Range | 5 to 100 ng |
Product Line | Quant-iT |
Shipping Condition | Room Temperature |
Unit Size | 1 kit |
Assay | RNA Quantitation, extended range |
Detection Method | Fluorescence |
For Use With (Equipment) | Microplate Reader |
Quantitation Range | 200 to 10,000 ng |
Product Line | Quant-iT |
Shipping Condition | Dry Ice |
Unit Size | 1000 assays |
Assay | RNA Quantitation, high sensitivity |
Detection Method | Fluorescence |
Excitation/Emission | 644/673 |
For Use With (Equipment) | Fluorometer, Microplate Reader |
No. of Reactions | 2000 Assays |
Quantitation Range | 5 to 100 ng |
Product Line | Quant-iT |
Shipping Condition | Room Temperature |
Unit Size | 1 mL |
Catalog Number | Specifications | Unit Size | Assay | Quantitation Range | Price (USD) | Availability | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
Q10213 | 1,000 assays | RNA Quantitation, broad range | 20 to 1000 ng | 523.65 Online price 562.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Q33140 | 1 kit | RNA Quantitation | 5 to 100 ng | 530.65 Online price 562.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Q33225 | 1000 assays | RNA Quantitation, extended range | 200 to 10,000 ng | 517.65 Online price 556.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Q32884 | 1 mL | RNA Quantitation, high sensitivity | 5 to 100 ng | 496.65 Online price 532.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|