Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Assay | RNA Quantitation, high sensitivity |
Detection Method | Fluorescence |
Excitation/Emission | 644/673 |
For Use With (Equipment) | Fluorometer, Microplate Reader |
No. of Reactions | 2000 Assays |
Quantitation Range | 5 to 100 ng |
Product Line | Quant-iT |
Shipping Condition | Room Temperature |
Unit Size | 1 mL |
Assay | RNA Quantitation |
Detection Method | Fluorescence |
For Use With (Equipment) | Microplate Reader |
No. of Reactions | 1000 Reactions (200 μL Assay Volume) |
Quantitation Range | 5 to 100 ng |
Product Line | Quant-iT |
Shipping Condition | Room Temperature |
Unit Size | 1 kit |
Assay | RNA Quantitation, broad range |
Detection Method | Fluorescence |
Excitation/Emission | 644/673 |
For Use With (Equipment) | Microplate Reader |
No. of Reactions | 1000 Reactions (200 μL Assay Volume) |
Quantitation Range | 20 to 1000 ng |
Product Line | Quant-iT |
Shipping Condition | Room Temperature |
Unit Size | 1,000 assays |
Assay | RNA Quantitation, extended range |
Detection Method | Fluorescence |
For Use With (Equipment) | Microplate Reader |
Quantitation Range | 200 to 10,000 ng |
Product Line | Quant-iT |
Shipping Condition | Dry Ice |
Unit Size | 1000 assays |
產品號碼 | 規格 | Unit Size | Assay | Quantitation Range | 價格 (TWD) | 供應狀況 | 數量 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
Q32884 | 1 mL | RNA Quantitation, high sensitivity | 5 to 100 ng | *** | *** | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Q33140 | 1 kit | RNA Quantitation | 5 to 100 ng | 價格 22,800.00 線上優惠 15,960.00 (01-Oct-2024 結束) 您的價格 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Q10213 | 1,000 assays | RNA Quantitation, broad range | 20 to 1000 ng | 價格 24,500.00 線上優惠 17,150.00 (01-Oct-2024 結束) 您的價格 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Q33225 | 1000 assays | RNA Quantitation, extended range | 200 to 10,000 ng | 與我們聯繫 › | - | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|