Invitrogen™

Qubit and Quant-it Assay Kit Standards

Catalog number: Q33236
Have Questions?
Invitrogen™

Qubit and Quant-it Assay Kit Standards

Catalog number: Q33236
Have Questions?
Achieve precise quantitation of dsDNA or RNA samples with stand-alone Qubit nucleic acid standards. The Qubit standards can be used in conjunction with different Qubit or Quant-iT nucleic acid quantitation assay kits and offer stand-alone versions of the kit standard for performing additional assays.
 
Catalog Number
Q33236
Unit Size
5 each
Product Type
Qubit RNA XR Standard
Shipping Condition
Dry Ice
Price (USD)
Availability
-
Full specifications
Quantity
Concentration8000 ng/μL
For Use With (Equipment)Qubit Fluorometer or fluorescence microplate reader
Product LineQubit
Product TypeQubit RNA XR Standard
Shipping ConditionDry Ice
Unit Size5 each
Catalog Number
Unit Size
5 mL
Product Type
1X dsDNA HS Assay Lambda Standard
Shipping Condition
Room Temperature
Price (USD)
Availability
-
Full specifications
Quantity
Concentration10 ng/μL
For Use With (Application)Qubit and Quant‐iT dsDNA HS Assays and 1 x dsDNA Assays
For Use With (Equipment)Qubit Fluorometer or fluorescence microplate reader
Green FeaturesFewer resources used, Less waste
Product LineQubit
Product Type1X dsDNA HS Assay Lambda Standard
Shipping ConditionRoom Temperature
Unit Size5 mL
Catalog Number
Unit Size
1 set
Product Type
Qubit RNA IQ Standards
Shipping Condition
Dry Ice
Price (USD)
Availability
-
Full specifications
Quantity
For Use With (Application)RNA Quantification
For Use With (Equipment)Qubit 4 Fluorometer, Qubit Flex Fluorometer
Product LineQubit
Product TypeQubit RNA IQ Standards
Shipping ConditionDry Ice
Unit Size1 set
Catalog Number
Unit Size
2 x 5 mL
Product Type
Qubit 1X dsDNA BR Lambda Standards
Shipping Condition
Wet Ice
Price (USD)
Availability
-
Full specifications
Quantity
Concentration0 ng/μL, 100 ng/μL
For Use With (Application)Qubit 1 x dsDNA BR Assay Kit or Qubit dsDNA BR Assay Kits
For Use With (Equipment)Qubit Fluorometer or fluorescence microplate reader
Green FeaturesFewer resources used, Less waste
Product LineQubit
Product TypeQubit 1X dsDNA BR Lambda Standards
Shipping ConditionWet Ice
Unit Size2 x 5 mL
Showing 4 of 4
Catalog NumberSpecificationsUnit SizeProduct TypeShipping ConditionPrice (USD)AvailabilityQuantity
Q33236Full specifications
5 eachQubit RNA XR StandardDry Ice
-
Concentration8000 ng/μL
For Use With (Equipment)Qubit Fluorometer or fluorescence microplate reader
Product LineQubit
Product TypeQubit RNA XR Standard
Shipping ConditionDry Ice
Unit Size5 each
Q33233Full specifications
5 mL1X dsDNA HS Assay Lambda StandardRoom Temperature
-
Concentration10 ng/μL
For Use With (Application)Qubit and Quant‐iT dsDNA HS Assays and 1 x dsDNA Assays
For Use With (Equipment)Qubit Fluorometer or fluorescence microplate reader
Green FeaturesFewer resources used, Less waste
Product LineQubit
Product Type1X dsDNA HS Assay Lambda Standard
Shipping ConditionRoom Temperature
Unit Size5 mL
Q33235Full specifications
1 setQubit RNA IQ StandardsDry Ice
-
For Use With (Application)RNA Quantification
For Use With (Equipment)Qubit 4 Fluorometer, Qubit Flex Fluorometer
Product LineQubit
Product TypeQubit RNA IQ Standards
Shipping ConditionDry Ice
Unit Size1 set
Q33263Full specifications
2 x 5 mLQubit 1X dsDNA BR Lambda StandardsWet Ice
-
Concentration0 ng/μL, 100 ng/μL
For Use With (Application)Qubit 1 x dsDNA BR Assay Kit or Qubit dsDNA BR Assay Kits
For Use With (Equipment)Qubit Fluorometer or fluorescence microplate reader
Green FeaturesFewer resources used, Less waste
Product LineQubit
Product TypeQubit 1X dsDNA BR Lambda Standards
Shipping ConditionWet Ice
Unit Size2 x 5 mL
Showing 4 of 4
Qubit 1X dsDNA HS Assay Lambda Standard
Qubit 1X dsDNA HS Assay Lambda Standard can be used with either Qubit 1X dsDNA HS Assay Kits, Qubit dsDNA HS Assay Kits, or their corresponding Quant-iT Assay Kits. The standards are supplied at the same concentration as the highest concentration dsDNA standard (10 ng/μL) in the Quant-iT Assay Kits and can be diluted into the standard series.

Qubit 1X dsDNA BR Standards
Qubit 1X dsDNA BR (Broad Range) Standards can be used with either Qubit 1X dsDNA BR Assay Kits, Qubit dsDNA BR Assay Kits, or their corresponding Quant-iT Assay Kits. The Qubit 1X dsDNA BR Standards includes standards at two concentrations, 0 and 100 ng/μL. The high standard corresponds to the same concentration as the highest concentration dsDNA standard in the Quant-iT Assay Kits and can be diluted into the standard series.

Qubit RNA XR Assay RNA Standard
Qubit RNA XR Standard can be used with either Qubit RNA XR Assay Kit or Quant-iT RNA XR Assay Kit. It is supplied at the same concentration as the highest concentration RNA standard in the Quant-iT RNA XR Assay Kit and can be diluted into the standard series. This standard cannot be used as a replacement for the large and/or structured standard in the Qubit RNA IQ Assay.

Qubit RNA IQ Assay Standards
Qubit RNA XR Standard can be used with either Qubit RNA XR Assay Kit or Quant-iT RNA XR Assay Kit. It is supplied at the same concentration as the highest concentration RNA standard (8000 ng/μL) in the Quant-iT RNA XR Assay Kit and can be diluted into the standard series. This standard cannot be used as a replacement for the large and/or structured standard in the Qubit RNA IQ Assay.

Note: Use thin-wall, clear, 0.5-mL PCR tubes (Cat. No. Q32856) for the Qubit 4 fluorometer and 8 x 200-μL tube strips (Cat. No. Q33252) for the Qubit Flex fluorometer.
For Research Use Only. Not for use in diagnostic procedures.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text