Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Concentration | 0 ng/μL, 100 ng/μL |
For Use With (Application) | Qubit 1 x dsDNA BR Assay Kit or Qubit dsDNA BR Assay Kits |
For Use With (Equipment) | Qubit Fluorometer or fluorescence microplate reader |
Green Features | Fewer resources used, Less waste |
Product Line | Qubit |
Product Type | Qubit 1X dsDNA BR Lambda Standards |
Shipping Condition | Wet Ice |
Unit Size | 2 x 5 mL |
Concentration | 10 ng/μL |
For Use With (Application) | Qubit and Quant‐iT dsDNA HS Assays and 1 x dsDNA Assays |
For Use With (Equipment) | Qubit Fluorometer or fluorescence microplate reader |
Green Features | Fewer resources used, Less waste |
Product Line | Qubit |
Product Type | 1X dsDNA HS Assay Lambda Standard |
Shipping Condition | Room Temperature |
Unit Size | 5 mL |
For Use With (Application) | RNA Quantification |
For Use With (Equipment) | Qubit 4 Fluorometer, Qubit Flex Fluorometer |
Product Line | Qubit |
Product Type | Qubit RNA IQ Standards |
Shipping Condition | Dry Ice |
Unit Size | 1 set |
Concentration | 8000 ng/μL |
For Use With (Equipment) | Qubit Fluorometer or fluorescence microplate reader |
Product Line | Qubit |
Product Type | Qubit RNA XR Standard |
Shipping Condition | Dry Ice |
Unit Size | 5 each |
Catalog Number | Specifications | Unit Size | Product Type | Shipping Condition | Price (USD) | Availability | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
Q33263 | 2 x 5 mL | Qubit 1X dsDNA BR Lambda Standards | Wet Ice | 96.65 Online price 98.50 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Q33233 | 5 mL | 1X dsDNA HS Assay Lambda Standard | Room Temperature | 78.65 Online price 80.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Q33235 | 1 set | Qubit RNA IQ Standards | Dry Ice | 77.65 Online price 79.75 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Q33236 | 5 each | Qubit RNA XR Standard | Dry Ice | 77.65 Online price 79.75 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|