Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Catalog Number | Quantity |
---|---|
QG0517 | 200 mL |
QS0518 | 20 mL |
QG0516 | 50 mL |
QuantiGene Homogenizing Solution is used to prepare tissue homogenates from fresh, frozen, or formalin-fixed paraffin-embedded (FFPE) tissues. Homogenization of tissues using QuantiGene Homogenizing Solution releases RNA for detection and quantitation with QuantiGene assays. The homogenizing solution is one of two components in the QuantiGene Sample Processing Kit for tissues, and is used to supplement this kit when extra volume is needed.
Share catalog number, name or link