QuantiGene Plex Wash Buffer Components are aqueous buffered solutions that are mixed with nuclease-free water to make QuantiGene Plex wash buffer. The wash buffer components may be purchased in 1, 3, or 10-plate quantities to supplement QuantiGene Plex Assay kits when additional wash buffer volume is required for automation.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Format
96-well
For Use With (Application)
QuantiGene Plex Assay
Key Functions
Multi-Gene Expression
Product Line
QuantiGene Plex
Product Type
Wash Buffer
Quantity
1 Plate
Shipping Condition
Room Temperature
Contents & Storage
• 0.8 mL Wash Buffer Component 1 • 13 mL Wash Buffer Component 2